Search
Search

RBL2| Gene Symbol: RBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323608.1 | NP_001310537.1 | 10 | 1569 | ||
| NM_001323609.1 | NP_001310538.1 | 10 | 1569 | |||
| NM_001323610.1 | NP_001310539.1 | 10 | 1569 | |||
| NM_001323611.1 | NP_001310540.1 | 10 | 1318 | |||
| NM_005611.3 | NP_005602.3 | 10 | 1550 | |||
| XM_011523253.2 | XP_011521555.1 | 8 | 943 | |||
| XM_017023513.1 | XP_016879002.1 | 7 | 1107 | |||
| GenBank mRNA | AK300047.1 | - | 10 | 1288 | ||
| AK304283.1 | - | 7 | 1017 | |||
| AK307881.1 | - | 10 | 1527 | |||
| BC034490.1 | - | 10 | 1540 | 4838444 | ||
| BX537767.1 | - | 9 | 1131 | |||
| S67171.1 | - | 10 | 1460 | |||
| X74594.1 | - | 10 | 1261 | |||
| X76061.1 | - | 10 | 1501 | |||