Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SAFB SIRNA Assays ›
          Gene Symbol:
          SAFB
          Gene Name:
          scaffold attachment factor B
          Gene Aliases:
          HAP, HET, SAB-B1, SAF-B, SAF-B1, SAFB1
          Chromosome Location:
          Chr.19: 5623035 - 5668478 on Build GRCh38
          UniGene ID:
          Hs.728802
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS109451
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SAFB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001201338.1 NP_001188267.1 13, 14 1921
          NM_001201339.1 NP_001188268.1 13, 14 1921
          NM_001201340.1 NP_001188269.1 12, 13 1714
          NM_001320571.1 NP_001307500.1 11, 12 1649
          NM_001320572.1 NP_001307501.1 13, 14 1921
          NM_002967.3 NP_002958.2 13, 14 1921
          XM_017027114.1 XP_016882603.1 11, 12 1603
          XM_017027115.1 XP_016882604.1 11, 12 1603
          XM_017027116.1 XP_016882605.1 12, 13 1311
          XM_017027117.1 XP_016882606.1 12, 13 1311
          XM_017027118.1 XP_016882607.1 12, 13 1311
          GenBank mRNA AB208780.1 - 13, 14 1838
          AK095736.1 - 9, 10 1097
          AK290444.1 - 13, 14 1817
          AK293152.1 - 10, 11 1419
          AK294671.1 - 11, 12 1374
          AK295298.1 - 10, 11 1507
          AK298707.1 - 12, 13 1610
          AK299525.1 - 13, 14 1682
          AK304474.1 - 9, 10 1072
          AK307970.1 - 13, 14 1821
          BC020309.1 - 13, 14 1803
          BC032055.1 - 13, 14 1800
          BC126219.1 - 13, 14 1792 8991935
          BC143937.1 - 13, 14 1789 9052455
          BC143938.1 - 13, 14 1792 9052456
          BC143939.1 - 13, 14 1792 9052457
          L43631.1 - 13, 14 1598
          U72355.1 - 13, 14 1803

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          transcription, DNA-templated
          intracellular estrogen receptor signaling pathway
          growth
          hormone metabolic process
          positive regulation of transcription from RNA polymerase II promoter
          regulation of mRNA processing
          nucleotide binding
          core promoter binding
          chromatin binding
          double-stranded DNA binding
          protein binding
          protein kinase binding
          sequence-specific DNA binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0