Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STIL SIRNA Assays ›
          Gene Symbol:
          STIL
          Gene Name:
          SCL/TAL1 interrupting locus
          Gene Aliases:
          MCPH7, SIL
          Chromosome Location:
          Chr.1: 47250139 - 47314787 on Build GRCh38
          UniGene ID:
          Hs.525198
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS109759
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STIL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001048166.1 NP_001041631.1 11, 12 1393
          NM_001282936.1 NP_001269865.1 12, 13 1602
          NM_001282937.1 NP_001269866.1 12, 13 1602
          NM_001282938.1 NP_001269867.1 13, 14 1461
          NM_001282939.1 NP_001269868.1 13, 14 1326
          NM_003035.2 NP_003026.2 11, 12 1393
          XM_006710834.3 XP_006710897.1 12, 13 1467
          XM_011541991.2 XP_011540293.1 12, 13 2133
          XM_011541992.2 XP_011540294.1 12, 13 2116
          XM_011541994.2 XP_011540296.1 11, 12 1393
          XM_011541996.2 XP_011540298.1 12, 13 1252
          XM_011541998.2 XP_011540300.1 12, 13 1252
          XM_011542001.2 XP_011540303.1 11, 12 1393
          XM_017002123.1 XP_016857612.1 12, 13 1252
          XM_017002124.1 XP_016857613.1 7, 8 669
          XM_017002125.1 XP_016857614.1 11, 12 1393
          XM_017002126.1 XP_016857615.1 11, 12 1393
          XM_017002127.1 XP_016857616.1 11, 12 1393
          GenBank mRNA AK128406.1 - 1, 2 1720
          AK307519.1 - 11, 12 1355
          BC053615.1 - 13, 14 1294
          BC126223.1 - 11, 12 1469 8991939
          BC144089.1 - 11, 12 1469 9052609
          CR749851.1 - 11, 12 1369
          M74558.1 - 12, 13 1618

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic axis specification
          in utero embryonic development
          neural tube closure
          heart looping
          mitotic spindle organization
          smoothened signaling pathway
          determination of left/right symmetry
          cell proliferation
          neural tube development
          forebrain development
          notochord development
          floor plate development
          multicellular organism growth
          negative regulation of apoptotic process
          regulation of centriole replication
          centrosome duplication
          protein localization to centrosome
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0