Search
Search

SLC2A1| Gene Symbol: SLC2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006516.2 | NP_006507.2 | 4 | 955 | ||
| GenBank mRNA | AB208987.1 | - | 4 | 675 | ||
| AK122999.1 | - | 2 | 215 | |||
| AK292791.1 | - | 4 | 641 | |||
| AK293306.1 | - | 4 | 638 | |||
| AK296736.1 | - | 5 | 542 | |||
| AK312403.1 | - | 4 | 647 | |||
| BC118590.1 | - | 4 | 576 | 40085221 | ||
| BC121804.1 | - | 4 | 576 | 40085220 | ||
| K03195.1 | - | 4 | 609 | |||