Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SVIL SIRNA Assays ›
Gene Symbol:
SVIL
Gene Name:
supervillin
Gene Aliases:
-
Chromosome Location:
Chr.10: 29457338 - 29737001 on Build GRCh38
UniGene ID:
Hs.499209
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS110392
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SVIL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001323599.1
NP_001310528.1
16
2611
NM_001323600.1
NP_001310529.1
14
2212
NM_003174.3
NP_003165.2
13
1997
NM_021738.2
NP_068506.2
15
3578
XM_005252570.2
XP_005252627.1
17
3475
XM_005252571.3
XP_005252628.1
17
3401
XM_005252573.2
XP_005252630.1
15
3607
XM_011519630.2
XP_011517932.2
15
4320
XM_011519631.2
XP_011517933.2
14
4224
XM_011519632.2
XP_011517934.2
14
4149
XM_011519633.2
XP_011517935.2
14
4224
XM_011519634.1
XP_011517936.1
17
3533
XM_011519635.1
XP_011517937.1
15
3500
XM_011519636.2
XP_011517938.2
14
3390
XM_011519637.2
XP_011517939.2
12
3138
XM_011519638.2
XP_011517940.2
11
3042
XM_017016575.1
XP_016872064.1
15
4320
XM_017016576.1
XP_016872065.1
14
4239
XM_017016577.1
XP_016872066.1
15
4320
XM_017016578.1
XP_016872067.1
13
3294
XM_017016579.1
XP_016872068.1
14
3390
XM_017016580.1
XP_016872069.1
13
3219
XM_017016581.1
XP_016872070.1
12
3138
XM_017016582.1
XP_016872071.1
11
3042
XM_017016583.1
XP_016872072.1
15
4320
GenBank mRNA
AF051850.1
-
13
1997
AF051851.1
-
9
1647
AF109135.1
-
15
3578
JX467682.1
-
10
2729
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
skeletal muscle tissue development
positive regulation of cytokinesis
protein binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0