Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TFDP2 SIRNA Assays ›
Gene Symbol:
TFDP2
Gene Name:
transcription factor Dp-2
Gene Aliases:
DP2
Chromosome Location:
Chr.3: 141944428 - 142149544 on Build GRCh38
UniGene ID:
Hs.379018
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS110661
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TFDP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178138.1
NP_001171609.1
8
973
NM_001178139.1
NP_001171610.1
7
938
NM_001178140.1
NP_001171611.1
4
434
NM_001178141.1
NP_001171612.1
4
367
NM_001178142.1
NP_001171613.1
2
215
NM_006286.4
NP_006277.1
5
489
XM_005247731.2
XP_005247788.1
7
938
XM_011513101.2
XP_011511403.1
8
1034
XM_011513102.2
XP_011511404.1
6
607
XM_011513106.2
XP_011511408.1
8
1034
XM_011513107.1
XP_011511409.1
7
938
XM_011513108.2
XP_011511410.1
5
761
XM_011513109.2
XP_011511411.1
3
333
XM_011513111.2
XP_011511413.1
3
333
XM_017007091.1
XP_016862580.1
7
949
XM_017007092.1
XP_016862581.1
10
1192
XM_017007093.1
XP_016862582.1
10
1381
XM_017007094.1
XP_016862583.1
7
967
XM_017007095.1
XP_016862584.1
9
1157
XM_017007096.1
XP_016862585.1
6
864
XM_017007097.1
XP_016862586.1
8
966
XM_017007098.1
XP_016862587.1
8
1111
XM_017007099.1
XP_016862588.1
9
1089
XM_017007100.1
XP_016862589.1
7
938
XM_017007101.1
XP_016862590.1
7
919
XM_017007102.1
XP_016862591.1
5
717
XM_017007103.1
XP_016862592.1
7
938
XM_017007104.1
XP_016862593.1
5
761
XM_017007105.1
XP_016862594.1
4
694
GenBank mRNA
AK303181.1
-
2
176
AK303634.1
-
4
309
AK314786.1
-
7
795
AL080206.1
-
4
394
BC021113.1
-
4
393
4653771
DC319314.1
-
5
489
L40386.1
-
4
333
U18422.1
-
5
441
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
protein binding
transcription factor binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline