Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TLE3 SIRNA Assays ›
          Gene Symbol:
          TLE3
          Gene Name:
          transducin like enhancer of split 3
          Gene Aliases:
          ESG, ESG3, GRG3, HsT18976
          Chromosome Location:
          Chr.15: 70047790 - 70097935 on Build GRCh38
          UniGene ID:
          Hs.287362
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS110791
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TLE3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105192.2 NP_001098662.1 3, 4 1300
          NM_001282979.1 NP_001269908.1 3, 4 1300
          NM_001282980.1 NP_001269909.1 3, 4 1300
          NM_001282981.1 NP_001269910.1 3, 4 233
          NM_005078.3 NP_005069.2 3, 4 1300
          NM_020908.2 NP_065959.1 3, 4 1300
          XM_005254622.4 XP_005254679.1 3, 4 1318
          XM_005254623.3 XP_005254680.1 3, 4 1318
          XM_005254625.3 XP_005254682.1 3, 4 1318
          XM_005254628.3 XP_005254685.1 3, 4 1318
          XM_005254633.2 XP_005254690.1 3, 4 1318
          XM_006720665.3 XP_006720728.1 3, 4 1318
          XM_011521976.2 XP_011520278.1 3, 4 1318
          XM_011521977.2 XP_011520279.1 3, 4 1318
          XM_011521978.2 XP_011520280.1 3, 4 1318
          XM_011521979.2 XP_011520281.1 2, 3 157
          XM_011521980.2 XP_011520282.1 3, 4 1318
          XM_011521981.1 XP_011520283.1 3, 4 1318
          XM_011521982.2 XP_011520284.1 3, 4 1318
          XM_011521983.2 XP_011520285.1 3, 4 1318
          XM_017022532.1 XP_016878021.1 3, 4 1318
          GenBank mRNA AB046767.1 - 3, 4 1282
          AK298482.1 - 3, 4 203
          AK315058.1 - 4, 5 628
          AY583459.1 - 3, 4 181
          AY583460.1 - 3, 4 181
          BC041831.1 - 2, 3 500 5268249
          BC043247.2 - 3, 4 1299 5296274
          M99438.1 - 3, 4 202

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053400, XM_006243216, XM_008766347, XM_006243220, XM_006243214, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001083928, NM_009389, NM_001083927, X73360, AK031322, and others.

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          signal transduction
          organ morphogenesis
          Wnt signaling pathway
          beta-catenin-TCF complex assembly
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0