Search
Search

TP53BP1| Gene Symbol: TP53BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141979.1 | NP_001135451.1 | 22 | 4948 | ||
| NM_001141980.1 | NP_001135452.1 | 22 | 4948 | |||
| NM_005657.2 | NP_005648.1 | 22 | 4968 | |||
| XM_011521985.1 | XP_011520287.1 | 21 | 4797 | |||
| XM_011521986.2 | XP_011520288.1 | 22 | 4947 | |||
| XM_017022533.1 | XP_016878022.1 | 23 | 5052 | |||
| GenBank mRNA | AB210025.1 | - | 22 | 4919 | ||
| AF078776.1 | - | 22 | 4993 | |||
| AK123418.1 | - | 1 | 954 | |||
| AK295764.1 | - | 22 | 4948 | |||
| BC112161.1 | - | 22 | 4928 | 8327629 | ||
| BX537418.1 | - | 22 | 4947 | |||
| U09477.1 | - | 9 | 1987 | |||