Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
TPM1
Gene Name:
tropomyosin 1 (alpha)
Gene Aliases:
C15orf13, CMD1Y, CMH3, HEL-S-265, HTM-alpha, LVNC9, TMSA
Chromosome Location:
Chr.15: 63042196 - 63071915 on Build GRCh38
UniGene ID:
Hs.133892
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS110926
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000366.5
NP_000357.3
3
445
NM_001018004.1
NP_001018004.1
3
445
NM_001018005.1
NP_001018005.1
3
445
NM_001018006.1
NP_001018006.1
3
445
NM_001018007.1
NP_001018007.1
3
445
NM_001018008.1
NP_001018008.1
2
285
NM_001018020.1
NP_001018020.1
3
445
NM_001301244.1
NP_001288173.1
3
445
NM_001301289.1
NP_001288218.1
2
285
XM_005254637.1
XP_005254694.1
4
526
XM_005254638.3
XP_005254695.2
3
888
XM_005254639.3
XP_005254696.2
3
888
XM_005254645.2
XP_005254702.2
3
888
XM_005254646.2
XP_005254703.1
2
271
XM_005254647.3
XP_005254704.1
2
271
XM_005254650.3
XP_005254707.1
2
271
XM_005254651.1
XP_005254708.1
2
271
XM_005254652.1
XP_005254709.1
2
271
XM_005254653.2
XP_005254710.1
2
271
XM_006720667.3
XP_006720730.2
3
888
XM_017022534.1
XP_016878023.1
3
888
XM_017022535.1
XP_016878024.1
3
400
XM_017022536.1
XP_016878025.1
3
888
XM_017022537.1
XP_016878026.1
3
400
XM_017022538.1
XP_016878027.1
3
888
XM_017022539.1
XP_016878028.1
3
888
XM_017022540.1
XP_016878029.1
2
271
XM_017022541.1
XP_016878030.1
2
271
GenBank mRNA
AB209041.1
-
2
322
AF474156.1
-
2
47
AK057989.1
-
4
293
AK092051.1
-
2
1162
AK092573.1
-
1
1267
AK129882.1
-
3
333
AK131384.1
-
4
459
AK298625.1
-
2
161
AK299387.1
-
3
334
AK301319.1
-
2
181
AK309170.1
-
3
654
AL050179.1
-
3
326
AY640414.1
-
3
254
AY640415.1
-
3
254
BC007433.2
-
3
395
2962925
BC050473.1
-
2
272
BC053545.1
-
2
283
5730451
BG107949.1
-
3
452
BX458841.2
-
3
314
CN423344.1
-
3
383
DQ424903.1
-
2
146
GQ472198.1
-
3
395
KU178366.1
-
3
254
KU178367.1
-
3
254
M19713.1
-
3
310
M19714.1
-
3
196
M19715.1
-
2
91
X12369.1
-
2
84
Z24727.1
-
469
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
movement of cell or subcellular component
muscle contraction
regulation of muscle contraction
cytoskeleton organization
regulation of heart contraction
regulation of cell shape
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
negative regulation of vascular smooth muscle cell proliferation
negative regulation of vascular associated smooth muscle cell migration
actin binding
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
structural constituent of muscle
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0