Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TSC1 SIRNA Assays ›
          Gene Symbol:
          TSC1
          Gene Name:
          tuberous sclerosis 1
          Gene Aliases:
          LAM, TSC
          Chromosome Location:
          Chr.9: 132891348 - 132945269 on Build GRCh38
          UniGene ID:
          Hs.370854
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS111008
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TSC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000368.4 NP_000359.1 7 759
          NM_001162426.1 NP_001155898.1 7 759
          NM_001162427.1 NP_001155899.1 6 606
          XM_005272211.1 XP_005272268.1 7 804
          XM_006717271.1 XP_006717334.1 7 739
          XM_011518979.2 XP_011517281.1 7 753
          XM_017015096.1 XP_016870585.1 8 1218
          XM_017015097.1 XP_016870586.1 8 1189
          XM_017015098.1 XP_016870587.1 7 804
          XM_017015099.1 XP_016870588.1 6 897
          XM_017015100.1 XP_016870589.1 6 868
          XM_017015101.1 XP_016870590.1 6 898
          GenBank mRNA AB190910.1 - 7 721
          AF013168.1 - 7 746
          AK297326.1 - 6 641
          AK299654.1 - 5 440
          AK303030.1 - 6 589
          AK308412.1 - 7 742
          BC047772.2 - 7 732
          BC070032.1 - 7 760
          BC108668.1 - 7 749
          BC121000.2 - 7 716
          KJ534953.1 - 5 525

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          neural tube closure
          regulation of cell-matrix adhesion
          adaptive immune response
          rRNA export from nucleus
          regulation of translation
          potassium ion transport
          cell cycle arrest
          cell-matrix adhesion
          negative regulation of cell proliferation
          adult locomotory behavior
          negative regulation of macroautophagy
          negative regulation of translation
          hippocampus development
          cerebral cortex development
          cell projection organization
          negative regulation of TOR signaling
          response to insulin
          myelination
          memory T cell differentiation
          regulation of phosphoprotein phosphatase activity
          negative regulation of cell size
          regulation of protein kinase activity
          glucose import
          negative regulation of insulin receptor signaling pathway
          synapse organization
          protein stabilization
          protein heterooligomerization
          regulation of stress fiber assembly
          regulation of cell cycle
          positive regulation of focal adhesion assembly
          cardiac muscle cell differentiation
          activation of GTPase activity
          cellular response to oxygen-glucose deprivation
          regulation of neuron death
          protein binding
          GTPase regulator activity
          GTPase activating protein binding
          protein N-terminus binding
          chaperone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0