Search
Search

HIRA| Gene Symbol: HIRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003325.3 | NP_003316.3 | 16 | 2013 | ||
| GenBank mRNA | AK289809.1 | - | 16 | 1793 | ||
| AK299950.1 | - | 17 | 1952 | |||
| AK302835.1 | - | 19 | 2239 | |||
| BC032721.1 | - | 16 | 2051 | |||
| BC039835.1 | - | 16 | 2008 | 5491361 | ||
| CR456503.1 | - | 16 | 1799 | |||
| X77633.1 | - | 15 | 1769 | |||
| X81844.1 | - | 16 | 1854 | |||
| X89887.1 | - | 16 | 2013 | |||