Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT1 SIRNA Assays ›
          Gene Symbol:
          WNT1
          Gene Name:
          Wnt family member 1
          Gene Aliases:
          BMND16, INT1, OI15
          Chromosome Location:
          Chr.12: 48978453 - 48982613 on Build GRCh38
          UniGene ID:
          Hs.248164
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS111347
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WNT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005430.3 NP_005421.1 2, 3 564
          GenBank mRNA BC074798.2 - 2, 3 402 30915522
          BC074799.2 - 2, 3 402 30915309
          BT019429.1 - 2, 3 366

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic axis specification
          branching involved in ureteric bud morphogenesis
          positive regulation of protein phosphorylation
          cell-cell signaling
          myoblast fusion
          positive regulation of cell proliferation
          response to wounding
          positive regulation of lamellipodium assembly
          negative regulation of cell-substrate adhesion
          Wnt signaling pathway
          spinal cord association neuron differentiation
          diencephalon development
          central nervous system morphogenesis
          cerebellum formation
          forebrain anterior/posterior pattern specification
          midbrain-hindbrain boundary maturation during brain development
          negative regulation of cell-cell adhesion
          negative regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of BMP signaling pathway
          ubiquitin-dependent SMAD protein catabolic process
          midbrain development
          organ regeneration
          T cell differentiation in thymus
          cell proliferation in midbrain
          inner ear morphogenesis
          signal transduction in response to DNA damage
          positive regulation of insulin-like growth factor receptor signaling pathway
          canonical Wnt signaling pathway involved in negative regulation of apoptotic process
          cell fate commitment
          negative regulation of fat cell differentiation
          positive regulation of Notch signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fibroblast proliferation
          neuron fate determination
          neuron development
          positive regulation of sequence-specific DNA binding transcription factor activity
          Spemann organizer formation
          canonical Wnt signaling pathway
          Wnt signaling pathway, planar cell polarity pathway
          bone development
          positive regulation of dermatome development
          hepatocyte differentiation
          cellular response to peptide hormone stimulus
          hematopoietic stem cell proliferation
          positive regulation of canonical Wnt signaling pathway
          negative regulation of cell aging
          beta-catenin destruction complex disassembly
          Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
          canonical Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
          embryonic brain development
          negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          frizzled binding
          cytokine activity
          morphogen activity
          protein domain specific binding
          transcription regulatory region DNA binding
          receptor agonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0