Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT5A SIRNA Assays ›
          Gene Symbol:
          WNT5A
          Gene Name:
          Wnt family member 5A
          Gene Aliases:
          hWNT5A
          Chromosome Location:
          Chr.3: 55465715 - 55505261 on Build GRCh38
          UniGene ID:
          Hs.643085
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS111356
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WNT5A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256105.1 NP_001243034.1 2, 3 200
          NM_003392.4 NP_003383.2 2, 3 795
          XM_006713324.1 XP_006713387.1 2, 3 155
          XM_011534085.2 XP_011532387.1 5, 6 5784
          XM_011534086.2 XP_011532388.1 5, 6 2408
          XM_011534087.2 XP_011532389.1 2, 3 301
          XM_011534088.2 XP_011532390.1 2, 3 1446
          XM_011534089.1 XP_011532391.1 2, 3 347
          XM_017007127.1 XP_016862616.1 6, 7 5511
          XM_017007128.1 XP_016862617.1 5, 6 2413
          GenBank mRNA AK290375.1 - 2, 3 458
          AK290869.1 - 2, 3 177
          BC064694.1 - 2, 3 456 30346200
          BC074783.2 - 1, 2 142
          DB224740.1 - 2, 3 200
          EF028086.1 - 2, 3 224
          L20861.1 - 1, 2 576

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          establishment of planar polarity
          somitogenesis
          epithelial to mesenchymal transition
          positive regulation of endothelial cell proliferation
          heart looping
          positive regulation of mesenchymal cell proliferation
          lens development in camera-type eye
          positive regulation of cytokine secretion involved in immune response
          type B pancreatic cell development
          planar cell polarity pathway involved in axis elongation
          optic cup formation involved in camera-type eye development
          Wnt signaling pathway, calcium modulating pathway
          activation of JUN kinase activity
          axon guidance
          synapse assembly
          hindgut morphogenesis
          midgut development
          male gonad development
          anterior/posterior axis specification, embryo
          response to organic substance
          positive regulation of endothelial cell migration
          positive regulation of peptidyl-threonine phosphorylation
          positive regulation of T cell chemotaxis
          positive regulation of neuron projection development
          Wnt signaling pathway
          olfactory bulb interneuron development
          neuron differentiation
          keratinocyte differentiation
          negative regulation of BMP signaling pathway
          positive regulation of cGMP metabolic process
          prostate gland development
          positive regulation of protein binding
          activation of protein kinase B activity
          response to estradiol
          positive regulation of interferon-gamma production
          positive regulation of interleukin-6 production
          positive regulation of peptidyl-serine phosphorylation
          response to testosterone
          cellular protein localization
          non-canonical Wnt signaling pathway
          post-anal tail morphogenesis
          chemoattraction of serotonergic neuron axon
          chemorepulsion of dopaminergic neuron axon
          non-canonical Wnt signaling pathway via JNK cascade
          negative regulation of fibroblast growth factor receptor signaling pathway
          wound healing
          embryonic digit morphogenesis
          positive regulation of macrophage activation
          negative regulation of apoptotic process
          estrous cycle
          positive regulation of chemokine biosynthetic process
          cell fate commitment
          establishment of epithelial cell apical/basal polarity
          negative regulation of fat cell differentiation
          positive regulation of protein catabolic process
          positive regulation of angiogenesis
          positive regulation of ossification
          positive regulation of endocytosis
          positive regulation of meiotic nuclear division
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of melanin biosynthetic process
          positive regulation of fibroblast proliferation
          lung alveolus development
          paraxial mesoderm formation
          notochord morphogenesis
          embryonic skeletal system development
          genitalia development
          negative regulation of axon extension involved in axon guidance
          hypophysis morphogenesis
          negative regulation of epithelial cell proliferation
          positive regulation of interleukin-1 beta secretion
          positive regulation of inflammatory response
          positive regulation of NF-kappaB transcription factor activity
          cartilage development
          response to glucocorticoid
          negative regulation of synapse assembly
          response to hyperoxia
          palate development
          convergent extension involved in axis elongation
          convergent extension involved in organogenesis
          uterus development
          cervix development
          vagina development
          canonical Wnt signaling pathway
          Wnt signaling pathway, planar cell polarity pathway
          urinary bladder development
          face development
          positive regulation of type I interferon-mediated signaling pathway
          lateral sprouting involved in mammary gland duct morphogenesis
          mesenchymal-epithelial cell signaling
          negative regulation of prostatic bud formation
          mammary gland branching involved in thelarche
          epithelial cell proliferation involved in mammary gland duct elongation
          positive regulation of response to cytokine stimulus
          regulation of branching involved in mammary gland duct morphogenesis
          planar cell polarity pathway involved in gastrula mediolateral intercalation
          mesodermal to mesenchymal transition involved in gastrulation
          positive regulation of macrophage cytokine production
          positive regulation of cartilage development
          planar cell polarity pathway involved in outflow tract morphogenesis
          planar cell polarity pathway involved in ventricular septum morphogenesis
          planar cell polarity pathway involved in cardiac right atrium morphogenesis
          planar cell polarity pathway involved in cardiac muscle tissue morphogenesis
          planar cell polarity pathway involved in pericardium morphogenesis
          positive regulation of thymocyte apoptotic process
          cellular response to lipopolysaccharide
          cellular response to calcium ion
          cellular response to retinoic acid
          cellular response to interferon-gamma
          hematopoietic stem cell proliferation
          cellular response to transforming growth factor beta stimulus
          negative regulation of mesenchymal cell proliferation
          primitive streak formation
          positive regulation of protein kinase C signaling
          negative regulation of canonical Wnt signaling pathway
          cochlea morphogenesis
          planar cell polarity pathway involved in neural tube closure
          activation of GTPase activity
          melanocyte proliferation
          positive regulation of protein kinase C activity
          positive regulation of protein localization to synapse
          regulation of cellular protein localization
          excitatory synapse assembly
          inhibitory synapse assembly
          negative regulation of cell proliferation in midbrain
          Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
          planar cell polarity pathway involved in midbrain dopaminergic neuron differentiation
          positive regulation of cell-cell adhesion mediated by cadherin
          positive regulation of non-canonical Wnt signaling pathway
          positive regulation of interleukin-8 secretion
          transcription factor activity, sequence-specific DNA binding
          frizzled binding
          frizzled-2 binding
          receptor tyrosine kinase-like orphan receptor binding
          cytokine activity
          protein binding
          protein domain specific binding
          transcription regulatory region DNA binding
          receptor agonist activity
          chemoattractant activity involved in axon guidance

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0