Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other YWHAZ SIRNA Assays ›
Gene Symbol:
YWHAZ
Gene Name:
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta
Gene Aliases:
14-3-3-zeta, HEL-S-3, HEL-S-93, HEL4, KCIP-1, YWHAD
Chromosome Location:
Chr.8: 100918576 - 100954068 on Build GRCh38
UniGene ID:
Hs.492407
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS111442
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: YWHAZ
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135699.1
NP_001129171.1
5
803
NM_001135700.1
NP_001129172.1
5
757
NM_001135701.1
NP_001129173.1
5
806
NM_001135702.1
NP_001129174.1
5
825
NM_003406.3
NP_003397.1
5
786
NM_145690.2
NP_663723.1
5
860
XM_005251061.3
XP_005251118.1
7
1183
XM_005251063.3
XP_005251120.1
5
955
XM_017013810.1
XP_016869299.1
6
972
XM_017013811.1
XP_016869300.1
7
1168
GenBank mRNA
AK289945.1
-
5
696
AK294647.1
-
4
445
AK296902.1
-
4
440
AK307506.1
-
6
663
AK315867.1
-
5
574
BC003623.2
-
5
753
2988020
BC013265.1
-
5
751
BC051814.1
-
5
874
BC063824.1
-
5
725
6180974
BC068456.1
-
5
774
4793291
BC072426.1
-
5
755
6315583
BC073141.1
-
5
709
BC083508.1
-
5
753
5563061
BC099904.1
-
5
775
5298373
BC101483.1
-
5
708
8068989
BC108281.1
-
5
763
3869494
BC111951.1
-
5
708
8327419
CR996247.1
-
5
758
DQ185048.1
-
2
232
EU794582.1
-
5
754
EU794583.1
-
5
774
GQ472215.1
-
5
774
M86400.1
-
5
732
U28964.1
-
5
774
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting
signal transduction
platelet activation
negative regulation of apoptotic process
regulation of mRNA stability
establishment of Golgi localization
membrane organization
Golgi reassembly
cell-cell adhesion
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
protein binding
transcription factor binding
protein kinase binding
protein domain specific binding
ubiquitin protein ligase binding
identical protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0