Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other YWHAZ SIRNA Assays ›
          Gene Symbol:
          YWHAZ
          Gene Name:
          tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta
          Gene Aliases:
          14-3-3-zeta, HEL-S-3, HEL-S-93, HEL4, KCIP-1, YWHAD
          Chromosome Location:
          Chr.8: 100918576 - 100954068 on Build GRCh38
          UniGene ID:
          Hs.492407
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS111443
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: YWHAZ
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135699.1 NP_001129171.1 5, 6 828
          NM_001135700.1 NP_001129172.1 5, 6 782
          NM_001135701.1 NP_001129173.1 5, 6 831
          NM_001135702.1 NP_001129174.1 5, 6 850
          NM_003406.3 NP_003397.1 5, 6 811
          NM_145690.2 NP_663723.1 5, 6 885
          XM_005251061.3 XP_005251118.1 7, 8 1208
          XM_005251063.3 XP_005251120.1 5, 6 980
          XM_017013810.1 XP_016869299.1 6, 7 997
          XM_017013811.1 XP_016869300.1 7, 8 1193
          GenBank mRNA AK289945.1 - 5, 6 721
          AK294647.1 - 4, 5 470
          AK296902.1 - 4, 5 465
          AK307506.1 - 6, 7 688
          AK315867.1 - 5, 6 599
          BC003623.2 - 5, 6 778 2988020
          BC013265.1 - 5, 6 776
          BC051814.1 - 5, 6 899
          BC063824.1 - 5, 6 750 6180974
          BC068456.1 - 5, 6 799 4793291
          BC072426.1 - 5, 6 780 6315583
          BC073141.1 - 5, 6 734
          BC083508.1 - 5, 6 778 5563061
          BC099904.1 - 5, 6 800 5298373
          BC101483.1 - 5, 6 733 8068989
          BC108281.1 - 5, 6 788 3869494
          BC111951.1 - 5, 6 733 8327419
          DQ185048.1 - 2, 3 257
          EU794582.1 - 5, 6 779
          EU794583.1 - 5, 6 799
          GQ472215.1 - 5, 6 799
          M86400.1 - 5, 6 757
          U28964.1 - 5, 6 799

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_013011, XM_006241529, D17615, L07913, U37252, and others.

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting
          signal transduction
          platelet activation
          negative regulation of apoptotic process
          regulation of mRNA stability
          establishment of Golgi localization
          membrane organization
          Golgi reassembly
          cell-cell adhesion
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          protein binding
          transcription factor binding
          protein kinase binding
          protein domain specific binding
          ubiquitin protein ligase binding
          identical protein binding
          poly(A) RNA binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0