Search
Search

HIST1H3I| Gene Symbol: HIST1H3I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003533.2 | NP_003524.1 | 1 | 179 | ||
| GenBank mRNA | BC066884.1 | - | 1 | 179 | 7002077 | |
| BC067493.1 | - | 1 | 179 | 7002079 | ||
| BC069305.1 | - | 1 | 179 | 7262745 | ||