Search
Search

PIP5K1B| Gene Symbol: PIP5K1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278253.1 | NP_001265182.1 | 4 | 549 | ||
| NM_003558.3 | NP_003549.1 | 5 | 706 | |||
| XM_005252261.3 | XP_005252318.1 | 4 | 746 | |||
| XM_005252262.4 | XP_005252319.1 | 5 | 502 | |||
| XM_006717300.2 | XP_006717363.1 | 5 | 612 | |||
| XM_006717301.1 | XP_006717364.1 | 5 | 423 | |||
| XM_011519082.2 | XP_011517384.1 | 5 | 3373 | |||
| XM_011519083.2 | XP_011517385.1 | 5 | 527 | |||
| XM_011519084.2 | XP_011517386.1 | 6 | 646 | |||
| XM_017015188.1 | XP_016870677.1 | 5 | 556 | |||
| XM_017015189.1 | XP_016870678.1 | 6 | 671 | |||
| XM_017015190.1 | XP_016870679.1 | 4 | 492 | |||
| XM_017015191.1 | XP_016870680.1 | 5 | 612 | |||
| GenBank mRNA | AK292734.1 | - | 5 | 423 | ||
| AK295587.1 | - | 4 | 304 | |||
| BC030587.2 | - | 6 | 540 | 4823087 | ||