Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA2 SIRNA Assays ›
Gene Symbol:
PPFIA2
Gene Name:
PTPRF interacting protein alpha 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 81257975 - 81759748 on Build GRCh38
UniGene ID:
Hs.506216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS112395
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220473.2
NP_001207402.1
6
923
NM_001220474.2
NP_001207403.1
5
869
NM_001220475.2
NP_001207404.1
6
735
NM_001220476.2
NP_001207405.1
6
735
NM_001220477.2
NP_001207406.1
4
742
NM_001282536.1
NP_001269465.1
4
430
NM_003625.4
NP_003616.2
7
786
XM_006719670.3
XP_006719733.1
4
7978
XM_011538906.2
XP_011537208.1
5
863
XM_011538907.2
XP_011537209.1
5
863
XM_017020079.1
XP_016875568.1
7
770
XM_017020080.1
XP_016875569.1
6
917
XM_017020081.1
XP_016875570.1
6
917
XM_017020082.1
XP_016875571.1
7
770
XM_017020083.1
XP_016875572.1
6
917
XM_017020084.1
XP_016875573.1
6
917
XM_017020085.1
XP_016875574.1
6
917
XM_017020086.1
XP_016875575.1
7
770
XM_017020087.1
XP_016875576.1
6
904
XM_017020088.1
XP_016875577.1
6
1150
XM_017020089.1
XP_016875578.1
5
863
XM_017020090.1
XP_016875579.1
7
770
XM_017020091.1
XP_016875580.1
6
917
XM_017020092.1
XP_016875581.1
6
917
XM_017020093.1
XP_016875582.1
6
917
XM_017020095.1
XP_016875584.1
6
917
XM_017020097.1
XP_016875586.1
7
770
XM_017020098.1
XP_016875587.1
7
770
XM_017020099.1
XP_016875588.1
5
863
XM_017020100.1
XP_016875589.1
6
917
XM_017020101.1
XP_016875590.1
7
770
XM_017020103.1
XP_016875592.1
6
917
XM_017020104.1
XP_016875593.1
6
917
XM_017020106.1
XP_016875595.1
6
917
XM_017020107.1
XP_016875596.1
7
770
XM_017020108.1
XP_016875597.1
7
770
XM_017020109.1
XP_016875598.1
6
917
XM_017020111.1
XP_016875600.1
6
917
XM_017020112.1
XP_016875601.1
4
742
XM_017020113.1
XP_016875602.1
4
742
XM_017020114.1
XP_016875603.1
4
742
XM_017020115.1
XP_016875604.1
4
742
XM_017020116.1
XP_016875605.1
4
742
XM_017020118.1
XP_016875607.1
5
717
XM_017020119.1
XP_016875608.1
4
742
XM_017020120.1
XP_016875609.1
4
430
GenBank mRNA
AB210009.1
-
6
921
AF034799.1
-
7
794
AK126971.1
-
5
829
AK297372.1
-
5
717
AK299853.1
-
4
691
AK302658.1
-
4
401
AK307524.1
-
7
770
BC104912.1
-
6
719
8143915
BC143485.1
-
6
719
9051993
BQ027545.1
-
1
148
DB083201.1
-
4
430
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
neurotransmitter secretion
glutamate secretion
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline