Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CADPS SIRNA Assays ›
          Gene Symbol:
          CADPS
          Gene Name:
          calcium dependent secretion activator
          Gene Aliases:
          CADPS1, CAPS, CAPS1, UNC-31
          Chromosome Location:
          Chr.3: 62398346 - 62875389 on Build GRCh38
          UniGene ID:
          Hs.654933
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS112615
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CADPS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003716.3 NP_003707.2 3 1069
          NM_183393.2 NP_899630.1 3 1069
          NM_183394.2 NP_899631.1 3 1069
          XM_006713378.2 XP_006713441.2 3 1064
          XM_011534177.1 XP_011532479.1 3 1064
          XM_011534178.1 XP_011532480.1 3 1064
          XM_011534180.1 XP_011532482.1 3 1064
          XM_011534182.1 XP_011532484.1 3 1064
          XM_011534183.1 XP_011532485.1 3 1064
          XM_011534189.1 XP_011532491.1 3 1064
          XM_011534191.2 XP_011532493.1 3 709
          XM_011534192.1 XP_011532494.1 3 1064
          XM_011534193.1 XP_011532495.1 3 1064
          XM_011534194.2 XP_011532496.1 3 709
          XM_011534195.1 XP_011532497.1 3 1064
          XM_011534196.1 XP_011532498.1 3 1064
          XM_011534197.2 XP_011532499.1 3 1022
          XM_011534198.2 XP_011532500.1 3 709
          XM_011534199.2 XP_011532501.1 3 709
          XM_011534200.2 XP_011532502.1 3 709
          XM_011534202.2 XP_011532504.1 3 1064
          XM_011534203.1 XP_011532505.1 3 1064
          XM_017007358.1 XP_016862847.1 3 1064
          XM_017007359.1 XP_016862848.1 3 1064
          XM_017007360.1 XP_016862849.1 3 1064
          XM_017007361.1 XP_016862850.1 3 1064
          XM_017007362.1 XP_016862851.1 3 1064
          XM_017007363.1 XP_016862852.1 3 1064
          XM_017007364.1 XP_016862853.1 3 1064
          XM_017007365.1 XP_016862854.1 3 1064
          XM_017007366.1 XP_016862855.1 3 1064
          XM_017007367.1 XP_016862856.1 3 709
          XM_017007368.1 XP_016862857.1 3 709
          XM_017007369.1 XP_016862858.1 3 1064
          XM_017007370.1 XP_016862859.1 3 1064
          XM_017007371.1 XP_016862860.1 3 1064
          XM_017007372.1 XP_016862861.1 3 709
          XM_017007373.1 XP_016862862.1 3 1064
          XM_017007374.1 XP_016862863.1 3 1064
          XM_017007375.1 XP_016862864.1 3 709
          XM_017007376.1 XP_016862865.1 3 1064
          XM_017007377.1 XP_016862866.1 3 709
          XM_017007378.1 XP_016862867.1 3 709
          XM_017007379.1 XP_016862868.1 3 1064
          XM_017007380.1 XP_016862869.1 3 1064
          XM_017007381.1 XP_016862870.1 3 1064
          XM_017007382.1 XP_016862871.1 3 709
          XM_017007383.1 XP_016862872.1 3 1064
          XM_017007384.1 XP_016862873.1 3 1064
          XM_017007385.1 XP_016862874.1 3 1064
          XM_017007386.1 XP_016862875.1 3 1064
          GenBank mRNA AB032947.2 - 3 1059
          AB593079.1 - 3 1022
          AF458662.1 - 3 1019
          AL833895.1 - 3 366

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          protein transport
          vesicle organization
          synaptic vesicle priming
          catecholamine secretion
          dense core granule exocytosis
          protein binding
          lipid binding
          protein kinase binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0