Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SGCE SIRNA Assays ›
          Gene Symbol:
          SGCE
          Gene Name:
          sarcoglycan epsilon
          Gene Aliases:
          DYT11, ESG
          Chromosome Location:
          Chr.7: 94585224 - 94656209 on Build GRCh38
          UniGene ID:
          Hs.371199
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS113191
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SGCE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099400.1 NP_001092870.1 3, 4 491
          NM_001099401.1 NP_001092871.1 3, 4 491
          NM_001301139.1 NP_001288068.1 2, 3 368
          NM_003919.2 NP_003910.1 3, 4 491
          XM_005250675.4 XP_005250732.1 4, 5 593
          XM_005250677.4 XP_005250734.1 4, 5 574
          XM_011516663.2 XP_011514965.1 4, 5 574
          XM_011516664.2 XP_011514966.1 4, 5 574
          XM_011516665.2 XP_011514967.1 3, 4 469
          XM_011516666.2 XP_011514968.1 3, 4 469
          XM_011516667.2 XP_011514969.1 5, 6 606
          XM_011516669.2 XP_011514971.1 4, 5 512
          XM_017012763.1 XP_016868252.1 4, 5 442
          XM_017012764.1 XP_016868253.1 3, 4 469
          XM_017012765.1 XP_016868254.1 5, 6 605
          XM_017012766.1 XP_016868255.1 4, 5 442
          XM_017012767.1 XP_016868256.1 4, 5 525
          XM_017012768.1 XP_016868257.1 4, 5 512
          XM_017012769.1 XP_016868258.1 4, 5 512
          XM_017012770.1 XP_016868259.1 4, 5 571
          GenBank mRNA AB117974.1 - 3, 4 343
          AF036364.1 - 3, 4 419
          AF516515.1 - 3, 4 380
          AJ000534.1 - 3, 4 448
          AK295021.1 - 4, 5 556
          AK298220.1 - 2, 3 325
          AK313438.1 - 3, 4 430
          AY359042.1 - 3, 4 397
          BC021709.2 - 3, 4 431 4822278
          BI552969.1 - 3, 4 496
          KU178449.1 - 3, 4 380
          KU178450.1 - 3, 4 380

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s)

          muscle system process
          cell-matrix adhesion
          muscle organ development
          membrane organization

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0