Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other P2RX6 SIRNA Assays ›
Gene Symbol:
P2RX6
Gene Name:
purinergic receptor P2X 6
Gene Aliases:
P2RXL1, P2X6, P2XM
Chromosome Location:
Chr.22: 21009699 - 21028013 on Build GRCh38
UniGene ID:
Hs.113275
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS113502
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: P2RX6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159554.1
NP_001153026.1
6, 7
580
NM_005446.3
NP_005437.2
6, 7
636
XM_005261819.3
XP_005261876.1
6, 7
1360
XM_011530498.2
XP_011528800.1
6, 7
1361
XM_011530499.2
XP_011528801.1
5, 6
1290
XM_011530500.2
XP_011528802.1
6, 7
1283
XM_011530501.2
XP_011528803.1
5, 6
628
XM_011530502.2
XP_011528804.1
6, 7
1361
XM_017029066.1
XP_016884555.1
6, 7
1361
XM_017029067.1
XP_016884556.1
5, 6
1288
XM_017029068.1
XP_016884557.1
6, 7
1360
XM_017029069.1
XP_016884558.1
5, 6
1209
XM_017029070.1
XP_016884559.1
5, 6
562
XM_017029071.1
XP_016884560.1
5, 6
1284
XM_017029072.1
XP_016884561.1
5, 6
517
XM_017029073.1
XP_016884562.1
5, 6
628
XM_017029074.1
XP_016884563.1
5, 6
481
XM_017029075.1
XP_016884564.1
5, 6
574
XM_017029076.1
XP_016884565.1
5, 6
1272
GenBank mRNA
AB002058.1
-
6, 7
651
AF065385.1
-
6, 7
650
BC033488.1
-
6, 7
620
5176046
BC047287.1
-
6, 7
619
5198937
BC063553.1
-
4, 5
473
BC064805.1
-
4, 5
432
BC109209.2
-
6, 7
563
40006731
BI753477.1
-
4, 5
432
CR456535.1
-
6, 7
642
Back To Top
More Information
Panther Classification:
Molecular Function -
ligand-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
transport
muscle contraction
signal transduction
blood coagulation
response to ATP
purinergic nucleotide receptor signaling pathway
protein homooligomerization
protein heterooligomerization
cation transmembrane transport
purinergic nucleotide receptor activity
transmembrane signaling receptor activity
extracellular ATP-gated cation channel activity
ATP binding
channel activity
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline