Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRFIP1 SIRNA Assays ›
Gene Symbol:
LRRFIP1
Gene Name:
LRR binding FLII interacting protein 1
Gene Aliases:
FLAP-1, FLAP1, FLIIAP1, GCF-2, GCF2, HUFI-1, TRIP
Chromosome Location:
Chr.2: 237627570 - 237781647 on Build GRCh38
UniGene ID:
Hs.471779
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS113634
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRFIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001137550.1
NP_001131022.1
19
1431
NM_001137551.1
NP_001131023.1
7
921
NM_001137552.1
NP_001131024.1
10
1089
NM_001137553.1
NP_001131025.1
7
921
NM_004735.3
NP_004726.2
9
1017
XM_005246141.4
XP_005246198.1
8
778
XM_005246142.2
XP_005246199.1
7
728
XM_017005253.1
XP_016860742.1
20
1493
XM_017005254.1
XP_016860743.1
20
1493
XM_017005255.1
XP_016860744.1
19
1421
XM_017005256.1
XP_016860745.1
18
1307
XM_017005257.1
XP_016860746.1
18
1358
XM_017005258.1
XP_016860747.1
18
1307
XM_017005259.1
XP_016860748.1
15
1220
XM_017005260.1
XP_016860749.1
15
1133
XM_017005261.1
XP_016860750.1
13
1070
XM_017005262.1
XP_016860751.1
13
1070
XM_017005263.1
XP_016860752.1
9
893
GenBank mRNA
AF115510.1
-
19
1300
AJ223075.1
-
9
903
AK298270.1
-
7
752
AK307485.1
-
10
819
BC001385.2
-
7
757
BC018883.2
-
7
940
BC019335.1
-
7
940
BC108913.1
-
7
746
40027210
BC108914.1
-
10
914
40027218
KP455483.1
-
7
629
U69609.1
-
7
753
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
biological_process
positive regulation of type I interferon production
negative regulation of transcription, DNA-templated
cell-cell adhesion
DNA binding
double-stranded RNA binding
protein binding
protein homodimerization activity
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0