Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRFIP2 SIRNA Assays ›
Gene Symbol:
LRRFIP2
Gene Name:
LRR binding FLII interacting protein 2
Gene Aliases:
HUFI-2
Chromosome Location:
Chr.3: 37052626 - 37176817 on Build GRCh38
UniGene ID:
Hs.740522
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS113635
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRFIP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134369.2
NP_001127841.1
9
1081
NM_001282691.1
NP_001269620.1
12
977
NM_006309.3
NP_006300.1
20
1476
NM_017724.2
NP_060194.1
8
1009
XM_005265539.2
XP_005265596.1
21
2476
XM_005265540.1
XP_005265597.1
22
1759
XM_005265550.1
XP_005265607.1
13
1312
XM_005265551.1
XP_005265608.1
12
1201
XM_005265553.1
XP_005265610.1
9
1048
XM_005265554.2
XP_005265611.1
9
1780
XM_005265556.1
XP_005265613.1
8
976
XM_006713385.1
XP_006713448.1
21
1741
XM_006713386.1
XP_006713449.1
20
1702
XM_006713387.1
XP_006713450.1
20
1699
XM_006713388.1
XP_006713451.1
20
1696
XM_006713389.1
XP_006713452.1
20
1693
XM_006713390.1
XP_006713453.1
21
1736
XM_006713392.1
XP_006713455.1
20
1672
XM_006713393.1
XP_006713456.1
20
1654
XM_006713394.1
XP_006713457.1
19
1651
XM_006713395.1
XP_006713458.1
21
1744
XM_006713396.1
XP_006713459.1
19
1636
XM_006713397.1
XP_006713460.1
18
1528
XM_006713400.1
XP_006713463.1
10
1093
XM_006713404.1
XP_006713467.1
9
1021
XM_011534217.1
XP_011532519.1
21
1744
XM_011534218.1
XP_011532520.1
22
1787
XM_011534219.1
XP_011532521.1
22
1855
XM_011534221.1
XP_011532523.1
19
1636
XM_011534222.1
XP_011532524.1
21
1744
XM_011534229.1
XP_011532531.1
11
1136
XM_017007465.1
XP_016862954.1
18
1591
XM_017007466.1
XP_016862955.1
19
1585
XM_017007467.1
XP_016862956.1
20
1672
XM_017007468.1
XP_016862957.1
16
1483
XM_017007469.1
XP_016862958.1
20
1672
XM_017007470.1
XP_016862959.1
17
1534
XM_017007471.1
XP_016862960.1
11
1150
XM_017007472.1
XP_016862961.1
12
1201
XM_017007473.1
XP_016862962.1
10
1078
XM_017007474.1
XP_016862963.1
11
1150
XM_017007475.1
XP_016862964.1
11
1882
XM_017007476.1
XP_016862965.1
10
1091
XM_017007477.1
XP_016862966.1
10
1810
XM_017007478.1
XP_016862967.1
9
1048
XM_017007479.1
XP_016862968.1
11
1150
XM_017007480.1
XP_016862969.1
9
1292
XM_017007481.1
XP_016862970.1
9
1048
XM_017007482.1
XP_016862971.1
8
976
XM_017007483.1
XP_016862972.1
9
1780
XM_017007484.1
XP_016862973.1
10
1091
GenBank mRNA
AF115509.1
-
20
1435
AK000255.1
-
7
694
AK024692.1
-
1
27
AK291514.1
-
9
1058
AK302282.1
-
12
951
BC053668.1
-
8
966
6146978
BX648794.1
-
5
460
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
biological_process
Wnt signaling pathway
protein binding
LRR domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0