Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRRFIP2 SIRNA Assays ›
          Gene Symbol:
          LRRFIP2
          Gene Name:
          LRR binding FLII interacting protein 2
          Gene Aliases:
          HUFI-2
          Chromosome Location:
          Chr.3: 37052626 - 37176817 on Build GRCh38
          UniGene ID:
          Hs.740522
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS113636
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LRRFIP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134369.2 NP_001127841.1 10, 11 1232
          NM_001282691.1 NP_001269620.1 14, 15 1221
          NM_006309.3 NP_006300.1 23, 24 1822
          NM_017724.2 NP_060194.1 9, 10 1160
          XM_005265539.2 XP_005265596.1 24, 25 2822
          XM_005265540.1 XP_005265597.1 25, 26 2105
          XM_005265550.1 XP_005265607.1 16, 17 1658
          XM_005265551.1 XP_005265608.1 15, 16 1547
          XM_005265553.1 XP_005265610.1 12, 13 1394
          XM_005265554.2 XP_005265611.1 12, 13 2126
          XM_005265556.1 XP_005265613.1 11, 12 1322
          XM_006713385.1 XP_006713448.1 24, 25 2087
          XM_006713386.1 XP_006713449.1 23, 24 2048
          XM_006713387.1 XP_006713450.1 23, 24 2045
          XM_006713388.1 XP_006713451.1 23, 24 2042
          XM_006713389.1 XP_006713452.1 23, 24 2039
          XM_006713390.1 XP_006713453.1 24, 25 2082
          XM_006713392.1 XP_006713455.1 23, 24 2018
          XM_006713393.1 XP_006713456.1 23, 24 2000
          XM_006713394.1 XP_006713457.1 22, 23 1997
          XM_006713395.1 XP_006713458.1 23, 24 1997
          XM_006713396.1 XP_006713459.1 22, 23 1982
          XM_006713397.1 XP_006713460.1 21, 22 1874
          XM_006713400.1 XP_006713463.1 13, 14 1439
          XM_006713404.1 XP_006713467.1 10, 11 1172
          XM_011534217.1 XP_011532519.1 24, 25 2090
          XM_011534218.1 XP_011532520.1 25, 26 2133
          XM_011534219.1 XP_011532521.1 25, 26 2201
          XM_011534221.1 XP_011532523.1 22, 23 1982
          XM_011534222.1 XP_011532524.1 22, 23 1895
          XM_011534229.1 XP_011532531.1 14, 15 1482
          XM_017007465.1 XP_016862954.1 21, 22 1937
          XM_017007466.1 XP_016862955.1 22, 23 1931
          XM_017007467.1 XP_016862956.1 22, 23 1925
          XM_017007468.1 XP_016862957.1 19, 20 1829
          XM_017007469.1 XP_016862958.1 21, 22 1823
          XM_017007470.1 XP_016862959.1 19, 20 1787
          XM_017007471.1 XP_016862960.1 14, 15 1496
          XM_017007472.1 XP_016862961.1 14, 15 1454
          XM_017007473.1 XP_016862962.1 13, 14 1424
          XM_017007474.1 XP_016862963.1 13, 14 1403
          XM_017007475.1 XP_016862964.1 13, 14 2135
          XM_017007476.1 XP_016862965.1 13, 14 1437
          XM_017007477.1 XP_016862966.1 12, 13 2063
          XM_017007478.1 XP_016862967.1 11, 12 1301
          XM_017007479.1 XP_016862968.1 12, 13 1301
          XM_017007480.1 XP_016862969.1 11, 12 1545
          XM_017007481.1 XP_016862970.1 11, 12 1292
          XM_017007482.1 XP_016862971.1 10, 11 1229
          XM_017007483.1 XP_016862972.1 10, 11 1931
          XM_017007484.1 XP_016862973.1 11, 12 1242
          GenBank mRNA AF115509.1 - 23, 24 1781
          AK000255.1 - 8, 9 845
          AK024692.1 - 2, 3 178
          AK124432.1 - 1, 2 2956
          AK291514.1 - 10, 11 1209
          AK302282.1 - 14, 15 1195
          BC053668.1 - 9, 10 1117 6146978
          BX648794.1 - 6, 7 611

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          biological_process
          Wnt signaling pathway
          protein binding
          LRR domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0