Search
Search

KIF23| Gene Symbol: KIF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281301.1 | NP_001268230.1 | 16 | 1996 | ||
| NM_004856.6 | NP_004847.2 | 17 | 2133 | |||
| NM_138555.3 | NP_612565.1 | 17 | 2133 | |||
| XM_005254796.2 | XP_005254853.1 | 18 | 2124 | |||
| XM_005254797.2 | XP_005254854.1 | 18 | 2124 | |||
| XM_005254798.2 | XP_005254855.1 | 18 | 2124 | |||
| XM_005254799.2 | XP_005254856.1 | 18 | 2124 | |||
| XM_011522238.1 | XP_011520540.1 | 17 | 1959 | |||
| XM_011522239.2 | XP_011520541.1 | 17 | 1984 | |||
| XM_011522240.1 | XP_011520542.1 | 9 | 978 | |||
| GenBank mRNA | AK303874.1 | - | 13 | 1527 | ||
| AK308322.1 | - | 16 | 1918 | |||
| AK308361.1 | - | 11 | 1342 | |||
| BC017705.2 | - | 16 | 1901 | |||
| BC051826.1 | - | 17 | 2075 | 6500787 | ||
| BX327654.2 | - | 2 | 128 | |||
| X67155.2 | - | 16 | 1973 | |||