Search
Search

KIF23| Gene Symbol: KIF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281301.1 | NP_001268230.1 | 6 | 795 | ||
| NM_004856.6 | NP_004847.2 | 7 | 932 | |||
| NM_138555.3 | NP_612565.1 | 7 | 932 | |||
| XM_005254796.2 | XP_005254853.1 | 7 | 881 | |||
| XM_005254797.2 | XP_005254854.1 | 7 | 881 | |||
| XM_005254798.2 | XP_005254855.1 | 7 | 881 | |||
| XM_005254799.2 | XP_005254856.1 | 7 | 881 | |||
| XM_011522238.1 | XP_011520540.1 | 6 | 716 | |||
| XM_011522239.2 | XP_011520541.1 | 7 | 783 | |||
| XM_017022733.1 | XP_016878222.1 | 7 | 881 | |||
| GenBank mRNA | AK303874.1 | - | 2 | 284 | ||
| AK308322.1 | - | 6 | 717 | |||
| AK308361.1 | - | 1 | 141 | |||
| AK310402.1 | - | 7 | 829 | |||
| BC017705.2 | - | 6 | 700 | |||
| BC051826.1 | - | 7 | 874 | 6500787 | ||
| X67155.2 | - | 6 | 772 | |||