Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other H2AFY SIRNA Assays ›
          Gene Symbol:
          H2AFY C5orf66
          Gene Name:
          H2A histone family member Y
          chromosome 5 open reading frame 66
          Gene Aliases:
          H2A.y, H2A/y, H2AF12M, MACROH2A1.1, mH2A1, macroH2A1.2
          -
          Chromosome Location:
          Chr.5: 135334381 - 135399887 on Build GRCh38
          Chromosome Location:
          Chr.5: 135033280 - 135344680 on Build GRCh38
          UniGene ID:
          Hs.420272
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS114261
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: H2AFY
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040158.1 NP_001035248.1 8 1025
          NM_004893.2 NP_004884.1 8 988
          NM_138609.2 NP_613075.1 8 1019
          NM_138610.2 NP_613258.2 8 1028
          XM_005272132.1 XP_005272189.1 8 1394
          XM_011543729.2 XP_011542031.1 8 945
          XM_011543730.2 XP_011542032.1 8 945
          GenBank mRNA AB209490.1 - 3 10086
          AF041483.1 - 8 925
          AF044286.1 - 7 811
          AF054174.1 - 8 988
          AK023409.1 - 8 1019
          AK123153.1 - 3 846
          AK296014.1 - 4 503
          AY134746.1 - 7 801
          BC013331.1 - 8 954 4077577
          BC095406.1 - 8 988 30348703
          Gene Symbol: C5orf66
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277348.1 NP_001264277.1 6 1185
          GenBank mRNA AK026965.1 - 6 1185

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001159515, NM_001159513, NM_001159514, NM_012015, AF171080, and others.

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of cell growth
          nucleosome assembly
          chromatin silencing
          dosage compensation
          covalent chromatin modification
          regulation of lipid metabolic process
          negative regulation of histone phosphorylation
          positive regulation of maintenance of mitotic sister chromatid cohesion
          regulation of gene expression, epigenetic
          positive regulation of keratinocyte differentiation
          negative regulation of gene expression, epigenetic
          positive regulation of gene expression, epigenetic
          negative regulation of histone H3-K4 methylation
          negative regulation of histone H3-K27 methylation
          regulation of chromatin silencing at rDNA
          establishment of protein localization to chromatin
          negative regulation of protein serine/threonine kinase activity
          negative regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter
          negative regulation of cell cycle G2/M phase transition
          negative regulation of protein localization to chromosome, telomeric region
          rDNA binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          DNA binding
          protein binding
          double-stranded methylated DNA binding
          enzyme binding
          protein kinase binding
          protein serine/threonine kinase inhibitor activity
          chromatin DNA binding
          nucleosomal DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0