Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DEPDC5 SIRNA Assays ›
Gene Symbol:
DEPDC5
Gene Name:
DEP domain containing 5
Gene Aliases:
DEP.5, FFEVF
Chromosome Location:
Chr.22: 31753951 - 31907034 on Build GRCh38
UniGene ID:
Hs.435022
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS114496
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DEPDC5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007188.2
NP_001007189.1
16, 17
1338
NM_001136029.2
NP_001129501.1
15, 16
1196
NM_001242896.1
NP_001229825.1
16, 17
1338
NM_001242897.1
NP_001229826.1
16, 17
1242
NM_014662.4
NP_055477.1
15, 16
1196
NR_110988.1
16, 17
1338
XM_005261862.1
XP_005261919.1
16, 17
1242
XM_011530557.2
XP_011528859.1
16, 17
1345
XM_011530558.2
XP_011528860.1
16, 17
1345
XM_011530559.2
XP_011528861.1
16, 17
1345
XM_011530560.2
XP_011528862.1
16, 17
1364
XM_011530561.2
XP_011528863.1
16, 17
1345
XM_011530562.2
XP_011528864.1
16, 17
1345
XM_011530563.2
XP_011528865.1
16, 17
1345
XM_011530565.2
XP_011528867.1
16, 17
1345
XM_011530568.2
XP_011528870.1
16, 17
1345
XM_017029109.1
XP_016884598.1
16, 17
1242
XM_017029110.1
XP_016884599.1
16, 17
1242
XM_017029111.1
XP_016884600.1
16, 17
1242
XM_017029112.1
XP_016884601.1
16, 17
1242
XM_017029113.1
XP_016884602.1
16, 17
1345
XM_017029114.1
XP_016884603.1
16, 17
1345
XM_017029115.1
XP_016884604.1
16, 17
1345
GenBank mRNA
AB014545.1
-
15, 16
1195
AJ698950.1
-
15, 16
1191
AJ698951.1
-
15, 16
1191
AJ704764.1
-
15, 16
1191
AK294987.1
-
16, 17
1214
AK300737.1
-
15, 16
1223
BC057797.1
-
16, 17
1278
BC136612.1
-
16, 17
1252
9020602
BC146766.1
-
15, 16
1195
9007209
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein modulator
protein-binding activity modulator
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of TOR signaling
cellular response to amino acid starvation
intracellular signal transduction
positive regulation of GTPase activity
negative regulation of TORC1 signaling
GTPase activator activity
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0