Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMG7 SIRNA Assays ›
Gene Symbol:
SMG7
Gene Name:
SMG7, nonsense mediated mRNA decay factor
Gene Aliases:
C1orf16, EST1C, SGA56M
Chromosome Location:
Chr.1: 183471962 - 183554193 on Build GRCh38
UniGene ID:
Hs.591463
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS114941
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMG7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174061.1
NP_001167532.1
20
3381
NM_173156.2
NP_775179.1
20
3263
NM_201568.2
NP_963862.1
20
3125
NM_201569.2
NP_963863.2
21
3275
XM_005245648.3
XP_005245705.1
23
4098
XM_005245649.3
XP_005245706.1
22
3961
XM_005245652.3
XP_005245709.1
21
3282
XM_005245653.4
XP_005245710.1
22
3384
XM_006711676.3
XP_006711739.1
22
3823
XM_006711678.3
XP_006711741.1
21
3144
XM_011510205.2
XP_011508507.1
24
4276
XM_011510206.2
XP_011508508.1
24
4138
XM_011510207.2
XP_011508509.1
22
4000
XM_011510208.2
XP_011508510.1
23
4126
XM_011510209.2
XP_011508511.1
23
3988
XM_011510210.2
XP_011508512.1
20
3114
XM_017002966.1
XP_016858455.1
23
3415
XM_017002967.1
XP_016858456.1
22
3273
XM_017002968.1
XP_016858457.1
19
2968
XM_017002969.1
XP_016858458.1
21
3475
XM_017002970.1
XP_016858459.1
21
3399
XM_017002971.1
XP_016858460.1
21
3337
GenBank mRNA
AB085674.1
-
20
3076
AK299178.1
-
20
3252
AL833675.1
-
8
1508
AY386363.1
-
13
2232
BC036381.1
-
21
3143
4829267
BC052565.1
-
13
2120
D87437.2
-
20
2969
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001005507, NM_001160257, NM_001160256, AK145908, BC082789, and others.
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
mRNA export from nucleus
regulation of dephosphorylation
protein binding
telomeric DNA binding
protein phosphatase 2A binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0