Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNAP91 SIRNA Assays ›
          Gene Symbol:
          SNAP91
          Gene Name:
          synaptosome associated protein 91
          Gene Aliases:
          AP180, CALM
          Chromosome Location:
          Chr.6: 83552885 - 83709408 on Build GRCh38
          UniGene ID:
          Hs.368046
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS114953
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNAP91
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242792.1 NP_001229721.1 22, 23 2318
          NM_001242793.1 NP_001229722.1 20, 21 2027
          NM_001256717.1 NP_001243646.1 22, 23 2385
          NM_001256718.1 NP_001243647.1 21, 22 2028
          NM_014841.2 NP_055656.1 22, 23 2321
          XM_005248770.4 XP_005248827.1 22, 23 2160
          XM_006715615.1 XP_006715678.1 22, 23 2112
          XM_011536265.1 XP_011534567.1 22, 23 2119
          XM_011536266.1 XP_011534568.1 22, 23 2231
          XM_011536269.1 XP_011534571.1 22, 23 2112
          XM_011536271.1 XP_011534573.1 21, 22 2070
          XM_011536273.2 XP_011534575.1 21, 22 2034
          XM_011536275.2 XP_011534577.1 21, 22 2028
          XM_011536276.2 XP_011534578.1 21, 22 2004
          XM_011536277.1 XP_011534579.1 24, 25 2400
          XM_017011554.1 XP_016867043.1 21, 22 2270
          XM_017011555.1 XP_016867044.1 21, 22 2154
          XM_017011556.1 XP_016867045.1 21, 22 2086
          XM_017011557.1 XP_016867046.1 22, 23 2289
          XM_017011558.1 XP_016867047.1 22, 23 2119
          XM_017011559.1 XP_016867048.1 22, 23 2218
          XM_017011560.1 XP_016867049.1 22, 23 2160
          XM_017011561.1 XP_016867050.1 22, 23 2231
          XM_017011562.1 XP_016867051.1 21, 22 2274
          XM_017011563.1 XP_016867052.1 20, 21 2268
          XM_017011564.1 XP_016867053.1 20, 21 2139
          XM_017011565.1 XP_016867054.1 20, 21 2197
          XM_017011566.1 XP_016867055.1 21, 22 2247
          XM_017011567.1 XP_016867056.1 21, 22 2050
          XM_017011568.1 XP_016867057.1 20, 21 2241
          XM_017011569.1 XP_016867058.1 21, 22 2247
          XM_017011570.1 XP_016867059.1 21, 22 2211
          XM_017011571.1 XP_016867060.1 21, 22 2082
          XM_017011572.1 XP_016867061.1 21, 22 2041
          XM_017011573.1 XP_016867062.1 21, 22 2140
          XM_017011574.1 XP_016867063.1 22, 23 2289
          XM_017011575.1 XP_016867064.1 21, 22 2205
          XM_017011576.1 XP_016867065.1 21, 22 2008
          XM_017011577.1 XP_016867066.1 21, 22 2134
          XM_017011578.1 XP_016867067.1 21, 22 2147
          XM_017011579.1 XP_016867068.1 21, 22 2205
          XM_017011580.1 XP_016867069.1 21, 22 2181
          XM_017011581.1 XP_016867070.1 21, 22 2011
          XM_017011582.1 XP_016867071.1 21, 22 2040
          XM_017011583.1 XP_016867072.1 21, 22 2168
          XM_017011584.1 XP_016867073.1 21, 22 2011
          XM_017011585.1 XP_016867074.1 21, 22 2052
          XM_017011586.1 XP_016867075.1 21, 22 2004
          XM_017011587.1 XP_016867076.1 21, 22 2110
          XM_017011588.1 XP_016867077.1 21, 22 2123
          XM_017011589.1 XP_016867078.1 20, 21 2139
          XM_017011590.1 XP_016867079.1 20, 21 1968
          GenBank mRNA AB014556.1 - 22, 23 2247
          AK289582.1 - 22, 23 2289
          AK294556.1 - 15, 16 1279
          AK294898.1 - 21, 22 2012
          CR749348.1 - 22, 23 2352

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          protein transport
          clathrin coat assembly
          regulation of clathrin-mediated endocytosis
          protein binding
          1-phosphatidylinositol binding
          protein kinase binding
          clathrin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0