Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNAP91 SIRNA Assays ›
Gene Symbol:
SNAP91
Gene Name:
synaptosome associated protein 91
Gene Aliases:
AP180, CALM
Chromosome Location:
Chr.6: 83552885 - 83709408 on Build GRCh38
UniGene ID:
Hs.368046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS114954
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNAP91
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242792.1
NP_001229721.1
27
2824
NM_001242793.1
NP_001229722.1
25
2533
NM_001242794.1
NP_001229723.1
17
1702
NM_001256717.1
NP_001243646.1
27
2891
NM_001256718.1
NP_001243647.1
26
2519
NM_014841.2
NP_055656.1
27
2827
XM_005248770.4
XP_005248827.1
27
2666
XM_006715615.1
XP_006715678.1
27
2618
XM_011536265.1
XP_011534567.1
27
2625
XM_011536266.1
XP_011534568.1
27
2737
XM_011536269.1
XP_011534571.1
27
2603
XM_011536271.1
XP_011534573.1
26
2576
XM_011536273.2
XP_011534575.1
26
2540
XM_011536275.2
XP_011534577.1
26
2534
XM_011536276.2
XP_011534578.1
26
2510
XM_011536277.1
XP_011534579.1
29
2906
XM_017011554.1
XP_016867043.1
26
2776
XM_017011555.1
XP_016867044.1
26
2660
XM_017011556.1
XP_016867045.1
26
2592
XM_017011557.1
XP_016867046.1
27
2780
XM_017011558.1
XP_016867047.1
27
2610
XM_017011559.1
XP_016867048.1
27
2709
XM_017011560.1
XP_016867049.1
27
2651
XM_017011561.1
XP_016867050.1
27
2722
XM_017011562.1
XP_016867051.1
26
2780
XM_017011563.1
XP_016867052.1
25
2774
XM_017011564.1
XP_016867053.1
25
2645
XM_017011565.1
XP_016867054.1
25
2703
XM_017011566.1
XP_016867055.1
26
2753
XM_017011567.1
XP_016867056.1
26
2556
XM_017011568.1
XP_016867057.1
25
2747
XM_017011569.1
XP_016867058.1
26
2738
XM_017011570.1
XP_016867059.1
26
2717
XM_017011571.1
XP_016867060.1
26
2588
XM_017011572.1
XP_016867061.1
26
2547
XM_017011573.1
XP_016867062.1
26
2646
XM_017011574.1
XP_016867063.1
27
2795
XM_017011575.1
XP_016867064.1
26
2711
XM_017011576.1
XP_016867065.1
26
2514
XM_017011577.1
XP_016867066.1
26
2640
XM_017011578.1
XP_016867067.1
26
2653
XM_017011579.1
XP_016867068.1
26
2696
XM_017011580.1
XP_016867069.1
26
2687
XM_017011581.1
XP_016867070.1
26
2517
XM_017011582.1
XP_016867071.1
26
2546
XM_017011583.1
XP_016867072.1
26
2659
XM_017011584.1
XP_016867073.1
26
2502
XM_017011585.1
XP_016867074.1
26
2543
XM_017011586.1
XP_016867075.1
26
2495
XM_017011587.1
XP_016867076.1
26
2601
XM_017011588.1
XP_016867077.1
26
2614
XM_017011589.1
XP_016867078.1
25
2645
XM_017011590.1
XP_016867079.1
25
2474
GenBank mRNA
AB014556.1
-
27
2753
AF054993.1
-
2
131
AK289582.1
-
27
2795
AK294556.1
-
20
1785
AK294898.1
-
26
2503
BC060818.1
-
17
1686
4827211
CR749348.1
-
27
2858
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
protein transport
clathrin coat assembly
regulation of clathrin-mediated endocytosis
protein binding
1-phosphatidylinositol binding
protein kinase binding
clathrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2w88v:80/100.66.78.250:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0