Search
Search

IRX5| Gene Symbol: IRX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252197.1 | NP_001239126.1 | 3 | 1393 | ||
| NM_005853.5 | NP_005844.4 | 3 | 1396 | |||
| XM_011522809.1 | XP_011521111.1 | 3 | 1839 | |||
| GenBank mRNA | AY335945.1 | - | 3 | 1393 | ||
| BC131510.1 | - | 3 | 738 | |||
| U90304.1 | - | 2 | 1573 | |||