Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ST3GAL6 SIRNA Assays ›
          Gene Symbol:
          ST3GAL6
          Gene Name:
          ST3 beta-galactoside alpha-2,3-sialyltransferase 6
          Gene Aliases:
          SIAT10, ST3GALVI
          Chromosome Location:
          Chr.3: 98732236 - 98795845 on Build GRCh38
          UniGene ID:
          Hs.148716
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS115927
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ST3GAL6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271142.1 NP_001258071.1 9 1027
          NM_001271145.1 NP_001258074.1 8 968
          NM_001271146.1 NP_001258075.1 8 957
          NM_001271147.1 NP_001258076.1 8 931
          NM_001271148.1 NP_001258077.1 5 715
          NM_001323352.1 NP_001310281.1 9 1129
          NM_001323353.1 NP_001310282.1 7 893
          NM_001323358.1 NP_001310287.1 10 1138
          NM_001323359.1 NP_001310288.1 9 1038
          NM_001323360.1 NP_001310289.1 10 1199
          NM_001323362.1 NP_001310291.1 8 1065
          NM_001323363.1 NP_001310292.1 7 904
          NM_001323364.1 NP_001310293.1 8 1076
          NM_001323365.1 NP_001310294.1 9 968
          NM_001323366.1 NP_001310295.1 10 1038
          NM_001323367.1 NP_001310296.1 8 785
          NM_001323368.1 NP_001310297.1 8 793
          NM_006100.3 NP_006091.1 9 1140
          GenBank mRNA AB022918.1 - 8 907
          AF119391.1 - 8 951
          AK001922.1 - 8 718
          AK315111.1 - 9 1131
          AL539506.3 - 5 572
          BC001266.2 - 9 955
          BC023312.1 - 9 1116 4996181

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          ganglioside biosynthetic process
          cellular protein modification process
          glycolipid metabolic process
          oligosaccharide metabolic process
          keratan sulfate biosynthetic process
          protein N-linked glycosylation via asparagine
          cellular response to interleukin-6
          sialylation
          beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
          beta-galactoside alpha-2,3-sialyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0