Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VAV3 SIRNA Assays ›
          Gene Symbol:
          VAV3
          Gene Name:
          vav guanine nucleotide exchange factor 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 107571160 - 107964923 on Build GRCh38
          UniGene ID:
          Hs.267659
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS116013
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: VAV3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001079874.1 NP_001073343.1 9, 10 905
          NM_006113.4 NP_006104.4 26, 27 2566
          XM_005270360.2 XP_005270417.1 25, 26 2378
          XM_017000053.1 XP_016855542.1 24, 25 2487
          XM_017000055.1 XP_016855544.1 25, 26 2236
          XM_017000056.1 XP_016855545.1 15, 16 1442
          GenBank mRNA AF067817.1 - 26, 27 2549
          AF118886.1 - 25, 26 2370
          AF118887.1 - 26, 27 2558
          AK296537.1 - 9, 10 1010
          AK300474.1 - 8, 9 860
          AK304088.1 - 9, 10 905
          AK316295.1 - 9, 10 905
          BC143969.1 - 27, 28 2733 9052487

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_146139, NM_020505, AK147053, BC027242, BC052739, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          vesicle fusion
          cellular response to DNA damage stimulus
          integrin-mediated signaling pathway
          small GTPase mediated signal transduction
          regulation of cell size
          positive regulation of signal transduction
          lamellipodium assembly
          platelet activation
          neutrophil chemotaxis
          positive regulation of B cell proliferation
          regulation of Rho protein signal transduction
          Fc-epsilon receptor signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          response to drug
          positive regulation of apoptotic process
          regulation of GTPase activity
          positive regulation of GTPase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of cell adhesion
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          B cell receptor signaling pathway
          regulation of small GTPase mediated signal transduction
          SH3/SH2 adaptor activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          GTPase activator activity
          epidermal growth factor receptor binding
          protein binding
          Rac guanyl-nucleotide exchange factor activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0