Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNCRIP SIRNA Assays ›
Gene Symbol:
SYNCRIP
Gene Name:
synaptotagmin binding cytoplasmic RNA interacting protein
Gene Aliases:
GRY-RBP, GRYRBP, HNRNPQ, HNRPQ1, NSAP1, PP68, hnRNP-Q
Chromosome Location:
Chr.6: 85607784 - 85643862 on Build GRCh38
UniGene ID:
Hs.571177
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS116098
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNCRIP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159673.1
NP_001153145.1
5, 6
634
NM_001159674.1
NP_001153146.1
6, 7
762
NM_001159675.1
NP_001153147.1
5, 6
668
NM_001159676.1
NP_001153148.1
5, 6
668
NM_001159677.1
NP_001153149.1
6, 7
1197
NM_001253771.1
NP_001240700.1
5, 6
576
NM_006372.4
NP_006363.4
6, 7
1197
XM_005248635.4
XP_005248692.1
6, 7
818
XM_005248636.2
XP_005248693.1
5, 6
1048
XM_005248637.2
XP_005248694.1
6, 7
781
XM_017010176.1
XP_016865665.1
6, 7
818
XM_017010177.1
XP_016865666.1
6, 7
818
XM_017010178.1
XP_016865667.1
6, 7
778
XM_017010179.1
XP_016865668.1
6, 7
753
XM_017010180.1
XP_016865669.1
5, 6
833
GenBank mRNA
AB209098.1
-
5, 6
574
AF037448.1
-
6, 7
1181
AF155568.1
-
6, 7
860
AK222776.1
-
6, 7
806
AK299796.1
-
5, 6
629
AK313572.1
-
6, 7
817
AY034481.1
-
5, 6
656
AY034482.1
-
5, 6
656
AY034483.1
-
5, 6
656
BC015575.2
-
6, 7
738
BC024283.1
-
6, 7
765
BC032643.1
-
6, 7
753
5495201
BC040844.1
-
5, 6
576
4513184
CV806885.1
-
2, 3
198
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
osteoblast differentiation
RNA processing
circadian rhythm
RNA splicing
viral process
negative regulation of translation
positive regulation of translation
CRD-mediated mRNA stabilization
cellular response to interferon-gamma
negative regulation of mRNA modification
nucleotide binding
RNA binding
mRNA 3'-UTR binding
protein binding
poly(A) binding
poly(A) RNA binding
mRNA 5'-UTR binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-9c8cf:80/100.66.75.150:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0