Search
Search

AHSA1| Gene Symbol: AHSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321441.1 | NP_001308370.1 | 6 | 650 | ||
| NM_012111.2 | NP_036243.1 | 6 | 784 | |||
| GenBank mRNA | AF161440.1 | - | 5 | 428 | ||
| AF164791.1 | - | 6 | 690 | |||
| AJ243310.1 | - | 6 | 755 | |||
| AK300766.1 | - | 6 | 782 | |||
| AK313824.1 | - | 6 | 718 | |||
| BC000321.2 | - | 6 | 732 | 2821759 | ||
| BC007398.2 | - | 5 | 396 | |||
| BQ073652.1 | - | 6 | 574 | |||
| CX786861.1 | - | 6 | 661 | |||