Search
Search

DCTN6| Gene Symbol: DCTN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006571.3 | NP_006562.1 | 4, 5 | 368 | ||
| GenBank mRNA | AK314128.1 | - | 4, 5 | 325 | ||
| BC013175.1 | - | 4, 5 | 312 | 3925231 | ||
| BI562535.1 | - | 4, 5 | 354 | |||
| CR456851.1 | - | 4, 5 | 281 | |||
| D84145.1 | - | 4, 5 | 368 | |||