Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SP140 SIRNA Assays ›
Gene Symbol:
SP140
Gene Name:
SP140 nuclear body protein
Gene Aliases:
LYSP100, LYSP100-A, LYSP100-B
Chromosome Location:
Chr.2: 230202924 - 230316571 on Build GRCh38
UniGene ID:
Hs.632549
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS117512
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SP140
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005176.2
NP_001005176.1
3
455
NM_001278451.1
NP_001265380.1
3
455
NM_001278452.1
NP_001265381.1
3
455
NM_001278453.1
NP_001265382.1
3
455
NM_007237.4
NP_009168.4
3
455
XM_005246252.2
XP_005246309.1
3
419
XM_005246253.2
XP_005246310.1
3
419
XM_005246254.2
XP_005246311.1
3
419
XM_005246255.2
XP_005246312.1
3
419
XM_005246256.2
XP_005246313.1
3
419
XM_006712223.3
XP_006712286.1
3
419
XM_011510515.1
XP_011508817.1
3
419
XM_011510516.1
XP_011508818.1
3
419
XM_011510517.2
XP_011508819.1
5
1283
XM_011510518.2
XP_011508820.1
4
555
XM_011510519.1
XP_011508821.1
3
419
XM_011510520.1
XP_011508822.1
3
419
XM_017003239.1
XP_016858728.1
3
419
XM_017003240.1
XP_016858729.1
3
419
XM_017003241.1
XP_016858730.1
3
419
XM_017003242.1
XP_016858731.1
3
419
XM_017003243.1
XP_016858732.1
3
419
XM_017003244.1
XP_016858733.1
4
555
XM_017003245.1
XP_016858734.1
3
419
XM_017003246.1
XP_016858735.1
3
419
XM_017003247.1
XP_016858736.1
3
419
XM_017003248.1
XP_016858737.1
3
419
XM_017003249.1
XP_016858738.1
3
419
XM_017003250.1
XP_016858739.1
3
419
XM_017003251.1
XP_016858740.1
4
562
XM_017003252.1
XP_016858741.1
3
419
GenBank mRNA
AA741304.1
-
2
396
AK301267.1
-
3
455
AK301317.1
-
3
456
BC038091.1
-
3
426
BC054890.1
-
3
425
BC070160.1
-
3
421
BC105743.1
-
3
438
40035747
BC105744.1
-
3
440
40035746
BC105960.1
-
3
440
40035752
DC391925.1
-
3
455
U36499.1
-
3
456
U36500.1
-
3
456
U63420.1
-
3
446
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
defense response
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0