Search
Search

CASC3| Gene Symbol: CASC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007359.4 | NP_031385.2 | 5 | 800 | ||
| XM_005257163.1 | XP_005257220.1 | 5 | 578 | |||
| GenBank mRNA | AK292425.1 | - | 5 | 739 | ||
| AK296682.1 | - | 5 | 741 | |||
| AK310635.1 | - | 5 | 746 | |||
| BC030008.1 | - | 5 | 535 | |||
| BC044656.1 | - | 5 | 683 | 5547561 | ||
| BC050526.1 | - | 5 | 703 | 5561860 | ||
| X80199.2 | - | 5 | 738 | |||