Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMS1 SIRNA Assays ›
Gene Symbol:
RIMS1
Gene Name:
regulating synaptic membrane exocytosis 1
Gene Aliases:
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location:
Chr.6: 71886703 - 72403143 on Build GRCh38
UniGene ID:
Hs.485729
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118007
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RIMS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001168407.1
NP_001161879.1
3
526
NM_001168408.1
NP_001161880.1
3
526
NM_001168409.1
NP_001161881.1
3
311
NM_001168410.1
NP_001161882.1
3
598
NM_014989.5
NP_055804.2
8
2113
XM_005248688.3
XP_005248745.1
8
2049
XM_011535604.2
XP_011533906.2
8
2049
XM_011535611.2
XP_011533913.2
8
2049
XM_011535615.2
XP_011533917.2
8
2049
XM_017010516.1
XP_016866005.1
8
2049
XM_017010517.1
XP_016866006.1
8
2049
XM_017010518.1
XP_016866007.1
8
2049
XM_017010519.1
XP_016866008.1
8
2049
XM_017010520.1
XP_016866009.1
8
2049
XM_017010521.1
XP_016866010.1
8
2049
XM_017010522.1
XP_016866011.1
8
2049
XM_017010523.1
XP_016866012.1
8
2049
XM_017010524.1
XP_016866013.1
8
2049
XM_017010525.1
XP_016866014.1
8
2049
XM_017010526.1
XP_016866015.1
8
2049
XM_017010527.1
XP_016866016.1
8
2049
XM_017010528.1
XP_016866017.1
8
2049
XM_017010529.1
XP_016866018.1
8
2049
XM_017010530.1
XP_016866019.1
8
2049
XM_017010531.1
XP_016866020.1
8
2049
XM_017010532.1
XP_016866021.1
8
2049
XM_017010533.1
XP_016866022.1
8
2049
XM_017010534.1
XP_016866023.1
3
622
XM_017010535.1
XP_016866024.1
3
622
XM_017010536.1
XP_016866025.1
3
619
XM_017010537.1
XP_016866026.1
3
619
XM_017010538.1
XP_016866027.1
3
727
XM_017010539.1
XP_016866028.1
3
614
XM_017010540.1
XP_016866029.1
3
613
XM_017010541.1
XP_016866030.1
3
613
XM_017010542.1
XP_016866031.1
3
613
XM_017010543.1
XP_016866032.1
3
532
XM_017010544.1
XP_016866033.1
3
615
XM_017010545.1
XP_016866034.1
3
612
XM_017010546.1
XP_016866035.1
3
725
XM_017010547.1
XP_016866036.1
3
725
XM_017010548.1
XP_016866037.1
3
610
XM_017010549.1
XP_016866038.1
3
610
XM_017010550.1
XP_016866039.1
3
993
XM_017010551.1
XP_016866040.1
3
993
XM_017010552.1
XP_016866041.1
3
850
XM_017010553.1
XP_016866042.1
3
850
XM_017010554.1
XP_016866043.1
3
846
XM_017010555.1
XP_016866044.1
3
846
GenBank mRNA
AB002338.1
-
9
1836
AB045726.1
-
9
2079
AB051866.1
-
9
2079
AK293828.1
-
3
201
AK294868.1
-
3
526
AK295001.1
-
3
444
AK295177.1
-
3
512
AK296303.1
-
3
367
AK308658.1
-
3
598
AK309185.1
-
3
311
AK311105.1
-
3
473
AK316108.1
-
3
324
AY190519.1
-
8
1792
BC151853.1
-
9
1860
8860502
BC152435.1
-
9
1836
8862556
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
intracellular protein transport
neurotransmitter secretion
visual perception
positive regulation of gene expression
glutamate secretion
synaptic vesicle exocytosis
calcium ion regulated exocytosis
regulated exocytosis
secretion
regulation of neurotransmitter secretion
regulation of catalytic activity
response to stimulus
long-term synaptic potentiation
membrane fusion
positive regulation of inhibitory postsynaptic potential
positive regulation of dendrite extension
positive regulation of excitatory postsynaptic potential
protein binding
SH3 domain binding
Rab GTPase binding
GTPase regulator activity
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0