Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMS1 SIRNA Assays ›
Gene Symbol:
RIMS1
Gene Name:
regulating synaptic membrane exocytosis 1
Gene Aliases:
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location:
Chr.6: 71886703 - 72403143 on Build GRCh38
UniGene ID:
Hs.485729
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118008
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RIMS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001168407.1
NP_001161879.1
14
1826
NM_001168408.1
NP_001161880.1
14
1829
NM_001168409.1
NP_001161881.1
14
1614
NM_001168410.1
NP_001161882.1
14
1901
NM_014989.5
NP_055804.2
19
3416
XM_005248688.3
XP_005248745.1
19
3352
XM_011535604.2
XP_011533906.2
19
3349
XM_011535615.2
XP_011533917.2
19
3349
XM_017010516.1
XP_016866005.1
19
3352
XM_017010517.1
XP_016866006.1
19
3352
XM_017010518.1
XP_016866007.1
19
3349
XM_017010519.1
XP_016866008.1
19
3352
XM_017010520.1
XP_016866009.1
19
3352
XM_017010521.1
XP_016866010.1
19
3352
XM_017010522.1
XP_016866011.1
19
3352
XM_017010523.1
XP_016866012.1
19
3352
XM_017010524.1
XP_016866013.1
19
3349
XM_017010525.1
XP_016866014.1
19
3352
XM_017010527.1
XP_016866016.1
19
3349
XM_017010530.1
XP_016866019.1
19
3352
XM_017010531.1
XP_016866020.1
19
3349
XM_017010532.1
XP_016866021.1
19
3352
XM_017010533.1
XP_016866022.1
19
3352
XM_017010534.1
XP_016866023.1
14
1925
XM_017010535.1
XP_016866024.1
14
1922
XM_017010536.1
XP_016866025.1
14
1922
XM_017010537.1
XP_016866026.1
14
1922
XM_017010538.1
XP_016866027.1
14
2030
XM_017010539.1
XP_016866028.1
14
1917
XM_017010540.1
XP_016866029.1
14
1916
XM_017010541.1
XP_016866030.1
14
1913
XM_017010542.1
XP_016866031.1
14
1916
XM_017010543.1
XP_016866032.1
14
1832
XM_017010545.1
XP_016866034.1
14
1912
XM_017010546.1
XP_016866035.1
14
2028
XM_017010547.1
XP_016866036.1
14
2025
XM_017010550.1
XP_016866039.1
14
2296
XM_017010551.1
XP_016866040.1
14
2293
XM_017010552.1
XP_016866041.1
14
2153
XM_017010553.1
XP_016866042.1
14
2150
XM_017010554.1
XP_016866043.1
14
2146
XM_017010555.1
XP_016866044.1
14
2146
XM_017010557.1
XP_016866046.1
2
152
XM_017010558.1
XP_016866047.1
1
45
GenBank mRNA
AB002338.1
-
20
3139
AB045726.1
-
20
3382
AB051866.1
-
20
3382
AF263305.1
-
7
770
AF263306.1
-
7
770
AF263307.1
-
7
767
AF263308.1
-
7
770
AF263309.1
-
7
767
AK293828.1
-
14
1504
AK294868.1
-
14
1826
AK295001.1
-
14
1747
AK296303.1
-
14
1670
AK300503.1
-
1
48
AK302571.1
-
4
367
AK308658.1
-
14
1901
AK309185.1
-
14
1614
AK316108.1
-
14
1627
AY190519.1
-
19
3095
BC151853.1
-
20
3163
8860502
BC152435.1
-
20
3139
8862556
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
intracellular protein transport
neurotransmitter secretion
visual perception
positive regulation of gene expression
glutamate secretion
synaptic vesicle exocytosis
calcium ion regulated exocytosis
regulated exocytosis
secretion
regulation of neurotransmitter secretion
regulation of catalytic activity
response to stimulus
long-term synaptic potentiation
membrane fusion
positive regulation of inhibitory postsynaptic potential
positive regulation of dendrite extension
positive regulation of excitatory postsynaptic potential
protein binding
SH3 domain binding
Rab GTPase binding
GTPase regulator activity
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0