Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HECW1 SIRNA Assays ›
Gene Symbol:
HECW1
Gene Name:
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases:
NEDL1
Chromosome Location:
Chr.7: 43112580 - 43566001 on Build GRCh38
UniGene ID:
Hs.164453
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118133
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HECW1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287059.1
NP_001273988.1
18
3725
NM_015052.4
NP_055867.3
20
4062
XM_005249665.3
XP_005249722.1
19
3844
XM_006715670.3
XP_006715733.1
18
3742
XM_006715671.3
XP_006715734.1
18
3739
XM_006715673.3
XP_006715736.1
17
3637
XM_011515220.2
XP_011513522.1
19
3635
XM_011515222.2
XP_011513524.1
18
3776
XM_011515223.2
XP_011513525.1
18
3577
XM_011515224.2
XP_011513526.1
18
4057
XM_011515225.2
XP_011513527.2
16
3094
XM_011515226.2
XP_011513528.1
15
3318
XM_017011882.1
XP_016867371.1
18
4084
XM_017011883.1
XP_016867372.1
19
4021
XM_017011884.1
XP_016867373.1
18
4081
XM_017011885.1
XP_016867374.1
20
4017
XM_017011886.1
XP_016867375.1
19
3846
XM_017011887.1
XP_016867376.1
17
3982
XM_017011888.1
XP_016867377.1
17
3979
XM_017011889.1
XP_016867378.1
18
4036
XM_017011890.1
XP_016867379.1
19
3846
GenBank mRNA
AB002320.2
-
20
4062
AB048365.1
-
17
3414
AK294918.1
-
18
3679
AL536075.3
-
4
357
BC151227.1
-
20
4062
9007236
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of canonical Wnt signaling pathway
ubiquitin-protein transferase activity
ligase activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline