Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41L3 SIRNA Assays ›
Gene Symbol:
EPB41L3
Gene Name:
erythrocyte membrane protein band 4.1 like 3
Gene Aliases:
4.1B, DAL-1, DAL1
Chromosome Location:
Chr.18: 5392381 - 5630666 on Build GRCh38
UniGene ID:
Hs.213394
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118235
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41L3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281533.1
NP_001268462.1
12
1664
NM_001281534.1
NP_001268463.1
12
1845
NM_001281535.1
NP_001268464.1
14
1646
NM_012307.3
NP_036439.2
12
1791
XM_011525623.2
XP_011523925.1
12
1845
XM_011525626.2
XP_011523928.1
11
1447
XM_011525628.2
XP_011523930.1
12
1845
XM_011525632.2
XP_011523934.1
11
1447
XM_011525634.2
XP_011523936.1
11
1447
XM_011525637.2
XP_011523939.1
11
1447
XM_017025618.1
XP_016881107.1
11
1447
XM_017025619.1
XP_016881108.1
11
1447
XM_017025620.1
XP_016881109.1
11
1447
XM_017025621.1
XP_016881110.1
11
1447
XM_017025622.1
XP_016881111.1
12
1791
XM_017025623.1
XP_016881112.1
11
1447
XM_017025624.1
XP_016881113.1
11
1447
XM_017025625.1
XP_016881114.1
12
1613
XM_017025626.1
XP_016881115.1
11
1447
XM_017025627.1
XP_016881116.1
12
1845
XM_017025628.1
XP_016881117.1
12
1845
XM_017025629.1
XP_016881118.1
12
1791
XM_017025630.1
XP_016881119.1
12
1559
XM_017025631.1
XP_016881120.1
11
1447
XM_017025632.1
XP_016881121.1
12
1845
XM_017025633.1
XP_016881122.1
12
1613
XM_017025634.1
XP_016881123.1
11
1447
XM_017025635.1
XP_016881124.1
11
1447
XM_017025636.1
XP_016881125.1
12
1845
XM_017025637.1
XP_016881126.1
12
1845
XM_017025638.1
XP_016881127.1
11
1447
XM_017025639.1
XP_016881128.1
12
1791
XM_017025640.1
XP_016881129.1
12
1845
XM_017025641.1
XP_016881130.1
11
1447
XM_017025642.1
XP_016881131.1
12
1791
XM_017025643.1
XP_016881132.1
12
1845
XM_017025644.1
XP_016881133.1
12
1791
XM_017025645.1
XP_016881134.1
12
1704
XM_017025646.1
XP_016881135.1
12
1845
XM_017025647.1
XP_016881136.1
12
1704
XM_017025648.1
XP_016881137.1
12
1704
XM_017025649.1
XP_016881138.1
12
1650
XM_017025650.1
XP_016881139.1
12
1704
XM_017025651.1
XP_016881140.1
12
1704
XM_017025652.1
XP_016881141.1
12
1650
XM_017025653.1
XP_016881142.1
12
1650
XM_017025654.1
XP_016881143.1
12
1704
XM_017025655.1
XP_016881144.1
12
1650
XM_017025656.1
XP_016881145.1
12
1704
XM_017025657.1
XP_016881146.1
12
1704
XM_017025658.1
XP_016881147.1
12
1650
XM_017025659.1
XP_016881148.1
12
1704
XM_017025660.1
XP_016881149.1
12
1650
GenBank mRNA
AB023204.1
-
12
1468
AF069072.1
-
12
1502
AK128531.1
-
13
1513
AK292583.1
-
14
1646
AK297406.1
-
12
1664
AK303679.1
-
2
127
AK307751.1
-
2
119
AK314438.1
-
10
1172
BC006141.1
-
12
1816
3936453
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013813, AF152247, AF177146, HG738870, AB523363, and others.
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
protein localization to paranode region of axon
apoptotic process
cytoskeletal anchoring at plasma membrane
biological_process
regulation of cell shape
cortical cytoskeleton organization
cortical actin cytoskeleton organization
paranodal junction assembly
myelin maintenance
neuron projection morphogenesis
protein localization to juxtaparanode region of axon
protein localization to plasma membrane
actin binding
structural constituent of cytoskeleton
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0