Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41L3 SIRNA Assays ›
Gene Symbol:
EPB41L3
Gene Name:
erythrocyte membrane protein band 4.1 like 3
Gene Aliases:
4.1B, DAL-1, DAL1
Chromosome Location:
Chr.18: 5392381 - 5630666 on Build GRCh38
UniGene ID:
Hs.213394
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118236
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41L3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281533.1
NP_001268462.1
4, 5
724
NM_001281534.1
NP_001268463.1
4, 5
905
NM_001281535.1
NP_001268464.1
6, 7
706
NM_012307.3
NP_036439.2
4, 5
905
XM_011525623.2
XP_011523925.1
4, 5
905
XM_011525626.2
XP_011523928.1
3, 4
507
XM_011525628.2
XP_011523930.1
4, 5
905
XM_011525632.2
XP_011523934.1
3, 4
507
XM_011525634.2
XP_011523936.1
3, 4
507
XM_011525637.2
XP_011523939.1
3, 4
507
XM_017025618.1
XP_016881107.1
3, 4
507
XM_017025619.1
XP_016881108.1
3, 4
507
XM_017025620.1
XP_016881109.1
3, 4
507
XM_017025621.1
XP_016881110.1
3, 4
507
XM_017025622.1
XP_016881111.1
4, 5
905
XM_017025623.1
XP_016881112.1
3, 4
507
XM_017025624.1
XP_016881113.1
3, 4
507
XM_017025625.1
XP_016881114.1
4, 5
673
XM_017025626.1
XP_016881115.1
3, 4
507
XM_017025627.1
XP_016881116.1
4, 5
905
XM_017025628.1
XP_016881117.1
4, 5
905
XM_017025629.1
XP_016881118.1
4, 5
905
XM_017025630.1
XP_016881119.1
4, 5
673
XM_017025631.1
XP_016881120.1
3, 4
507
XM_017025632.1
XP_016881121.1
4, 5
905
XM_017025633.1
XP_016881122.1
4, 5
673
XM_017025634.1
XP_016881123.1
3, 4
507
XM_017025635.1
XP_016881124.1
3, 4
507
XM_017025636.1
XP_016881125.1
4, 5
905
XM_017025637.1
XP_016881126.1
4, 5
905
XM_017025638.1
XP_016881127.1
3, 4
507
XM_017025639.1
XP_016881128.1
4, 5
905
XM_017025640.1
XP_016881129.1
4, 5
905
XM_017025641.1
XP_016881130.1
3, 4
507
XM_017025642.1
XP_016881131.1
4, 5
905
XM_017025643.1
XP_016881132.1
4, 5
905
XM_017025644.1
XP_016881133.1
4, 5
905
XM_017025645.1
XP_016881134.1
4, 5
764
XM_017025646.1
XP_016881135.1
4, 5
905
XM_017025647.1
XP_016881136.1
4, 5
764
XM_017025648.1
XP_016881137.1
4, 5
764
XM_017025649.1
XP_016881138.1
4, 5
764
XM_017025650.1
XP_016881139.1
4, 5
764
XM_017025651.1
XP_016881140.1
4, 5
764
XM_017025652.1
XP_016881141.1
4, 5
764
XM_017025653.1
XP_016881142.1
4, 5
764
XM_017025654.1
XP_016881143.1
4, 5
764
XM_017025655.1
XP_016881144.1
4, 5
764
XM_017025656.1
XP_016881145.1
4, 5
764
XM_017025657.1
XP_016881146.1
4, 5
764
XM_017025658.1
XP_016881147.1
4, 5
764
XM_017025659.1
XP_016881148.1
4, 5
764
XM_017025660.1
XP_016881149.1
4, 5
764
GenBank mRNA
AB023204.1
-
4, 5
582
AF069072.1
-
4, 5
562
AK128531.1
-
5, 6
573
AK292583.1
-
6, 7
706
AK307904.1
-
4, 5
826
AK314438.1
-
2, 3
232
BC006141.1
-
4, 5
876
3936453
BC008377.1
-
4, 5
741
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
protein localization to paranode region of axon
apoptotic process
cytoskeletal anchoring at plasma membrane
biological_process
regulation of cell shape
cortical cytoskeleton organization
cortical actin cytoskeleton organization
paranodal junction assembly
myelin maintenance
neuron projection morphogenesis
protein localization to juxtaparanode region of axon
protein localization to plasma membrane
actin binding
structural constituent of cytoskeleton
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0