Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PASK SIRNA Assays ›
Gene Symbol:
PASK
Gene Name:
PAS domain containing serine/threonine kinase
Gene Aliases:
PASKIN, STK37
Chromosome Location:
Chr.2: 241106099 - 241150347 on Build GRCh38
UniGene ID:
Hs.397891
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118314
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PASK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001252119.1
NP_001239048.1
10
2393
NM_001252120.1
NP_001239049.1
11
2546
NM_001252122.1
NP_001239051.1
10
2288
NM_001252124.1
NP_001239053.1
10
2393
NM_015148.3
NP_055963.2
10
2393
XM_005246991.1
XP_005247048.1
7
1728
XM_011510826.1
XP_011509128.1
10
2348
XM_011510828.1
XP_011509130.1
11
2449
XM_011510829.1
XP_011509131.1
10
2390
XM_011510830.1
XP_011509132.1
10
2391
XM_011510831.2
XP_011509133.1
8
2555
XM_011510832.2
XP_011509134.1
8
2555
XM_011510833.1
XP_011509135.1
7
1728
XM_011510834.1
XP_011509136.1
3
1451
XM_011510835.1
XP_011509137.1
3
1041
XM_017003634.1
XP_016859123.1
10
2637
XM_017003635.1
XP_016859124.1
11
2449
XM_017003636.1
XP_016859125.1
10
2405
GenBank mRNA
AF387103.1
-
10
2333
AK056516.1
-
10
2338
AK296945.1
-
7
1898
AK302417.1
-
10
2244
AK302561.1
-
7
1728
AL117400.1
-
2
2692
BC020054.1
-
6
1568
BC043495.1
-
4
1150
BC050565.1
-
11
2352
4537404
BC063585.1
-
10
2336
4419998
CR749231.1
-
11
2547
D50925.2
-
10
2324
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
regulation of respiratory gaseous exchange
negative regulation of glycogen biosynthetic process
positive regulation of translation
protein autophosphorylation
regulation of glucagon secretion
regulation of energy homeostasis
protein serine/threonine kinase activity
protein binding
ATP binding
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline