Search
Search

MTCL1| Gene Symbol: MTCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015210.3 | NP_056025.2 | 9, 10 | 2375 | ||
| XM_005258098.3 | XP_005258155.1 | 7, 8 | 3275 | |||
| XM_005258099.4 | XP_005258156.1 | 7, 8 | 3275 | |||
| XM_011525640.2 | XP_011523942.1 | 9, 10 | 3476 | |||
| XM_011525641.2 | XP_011523943.1 | 8, 9 | 3398 | |||
| XM_017025671.1 | XP_016881160.1 | 9, 10 | 2845 | |||
| XM_017025672.1 | XP_016881161.1 | 9, 10 | 2446 | |||
| XM_017025673.1 | XP_016881162.1 | 10, 11 | 2453 | |||
| XM_017025674.1 | XP_016881163.1 | 9, 10 | 2500 | |||
| XM_017025675.1 | XP_016881164.1 | 9, 10 | 2375 | |||
| GenBank mRNA | AB018345.2 | - | 7, 8 | 2292 | ||
| AK131528.1 | - | 9, 10 | 2375 | |||
| BC040542.1 | - | 2, 3 | 272 | 5268201 | ||