Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GPR161 SIRNA Assays ›
Gene Symbol:
GPR161
Gene Name:
G protein-coupled receptor 161
Gene Aliases:
RE2
Chromosome Location:
Chr.1: 168079542 - 168137667 on Build GRCh38
UniGene ID:
Hs.271809
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118761
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GPR161
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001267609.1
NP_001254538.1
4
1446
NM_001267610.1
NP_001254539.1
4
1409
NM_001267611.1
NP_001254540.1
3
996
NM_001267612.1
NP_001254541.1
3
952
NM_001267613.1
NP_001254542.1
2
845
NM_001267614.1
NP_001254543.1
2
578
NM_153832.2
NP_722561.1
5
1403
XM_005245055.2
XP_005245112.1
4
1370
XM_005245056.2
XP_005245113.1
3
977
XM_005245057.4
XP_005245114.1
3
994
XM_006711251.1
XP_006711314.1
5
1416
XM_006711253.2
XP_006711316.2
3
1209
XM_011509370.1
XP_011507672.1
5
1714
XM_011509371.2
XP_011507673.1
5
1160
XM_011509372.1
XP_011507674.1
5
1492
XM_011509373.2
XP_011507675.1
5
1418
XM_011509375.2
XP_011507677.1
5
1204
XM_011509376.2
XP_011507678.1
4
1335
XM_011509377.1
XP_011507679.1
3
1291
XM_011509378.1
XP_011507680.1
3
1263
XM_017000858.1
XP_016856347.1
4
1863
XM_017000859.1
XP_016856348.1
3
1817
XM_017000860.1
XP_016856349.1
4
1911
XM_017000861.1
XP_016856350.1
5
1957
XM_017000862.1
XP_016856351.1
5
1540
GenBank mRNA
AF091890.1
-
2
828
AK056040.1
-
3
899
AK122656.1
-
4
1325
AK298797.1
-
2
803
AK298850.1
-
2
578
AK299634.1
-
4
1077
AK310019.1
-
3
913
AY275468.1
-
2
768
BC028163.1
-
5
1082
5246359
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
multicellular organism development
positive regulation of cAMP biosynthetic process
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
G-protein coupled receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0