Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PNISR SIRNA Assays ›
Gene Symbol:
PNISR
Gene Name:
PNN interacting serine and arginine rich protein
Gene Aliases:
C6orf111, HSPC306, SFRS18, SRrp130, bA98I9.2
Chromosome Location:
Chr.6: 99398051 - 99425345 on Build GRCh38
UniGene ID:
Hs.520287
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS119580
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PNISR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001322405.1
NP_001309334.1
6, 7
905
NM_001322406.1
NP_001309335.1
7, 8
1021
NM_001322408.1
NP_001309337.1
5, 6
825
NM_001322410.1
NP_001309339.1
6, 7
909
NM_001322412.1
NP_001309341.1
6, 7
905
NM_001322413.1
NP_001309342.1
5, 6
829
NM_001322414.1
NP_001309343.1
6, 7
909
NM_001322415.1
NP_001309344.1
5, 6
829
NM_001322416.1
NP_001309345.1
6, 7
909
NM_015491.2
NP_056306.1
5, 6
829
NM_032870.3
NP_116259.2
6, 7
909
NR_136326.1
5, 6
829
XM_005266912.3
XP_005266969.2
5, 6
788
XM_017010710.1
XP_016866199.1
6, 7
906
XM_017010711.1
XP_016866200.1
5, 6
792
XM_017010712.1
XP_016866201.1
6, 7
2260
XM_017010713.1
XP_016866202.1
6, 7
2256
GenBank mRNA
AF314184.1
-
4, 5
699
AF314186.1
-
6, 7
847
AK027658.1
-
6, 7
872
AK027751.1
-
1, 2
1814
AK027759.1
-
7, 8
848
AK027898.1
-
6, 7
865
AK074628.1
-
5, 6
792
AK291155.1
-
4, 5
678
BC007791.2
-
6, 7
842
BC012780.2
-
6, 7
851
BC019074.2
-
6, 7
818
BC024655.1
-
1, 2
273
BC042850.1
-
7, 8
1018
BC052638.1
-
6, 7
847
BC064413.1
-
6, 7
843
BC064640.1
-
6, 7
822
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mfv9s:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0