Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SIPA1L1 SIRNA Assays ›
Gene Symbol:
SIPA1L1
Gene Name:
signal induced proliferation associated 1 like 1
Gene Aliases:
E6TP1
Chromosome Location:
Chr.14: 71320418 - 71741229 on Build GRCh38
UniGene ID:
Hs.654657
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS119717
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SIPA1L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284245.1
NP_001271174.1
2
1596
NM_001284246.1
NP_001271175.1
2
2185
NM_001284247.1
NP_001271176.1
1
1537
NM_015556.2
NP_056371.1
2
1596
XM_005267516.4
XP_005267573.1
2
2185
XM_006720111.2
XP_006720174.1
4
3475
XM_011536630.1
XP_011534932.1
6
1947
XM_011536631.1
XP_011534933.1
6
2358
XM_011536632.1
XP_011534934.1
5
1867
XM_011536635.2
XP_011534937.1
5
2014
XM_017021159.1
XP_016876648.1
8
2104
XM_017021160.1
XP_016876649.1
6
2026
XM_017021161.1
XP_016876650.1
7
1979
XM_017021162.1
XP_016876651.1
9
2161
XM_017021163.1
XP_016876652.1
4
1828
XM_017021164.1
XP_016876653.1
4
1835
XM_017021165.1
XP_016876654.1
6
1904
XM_017021166.1
XP_016876655.1
7
2037
XM_017021167.1
XP_016876656.1
6
1912
XM_017021168.1
XP_016876657.1
6
1886
XM_017021169.1
XP_016876658.1
5
2039
XM_017021170.1
XP_016876659.1
5
2397
XM_017021171.1
XP_016876660.1
6
1970
XM_017021172.1
XP_016876661.1
5
1845
XM_017021173.1
XP_016876662.1
4
1972
XM_017021174.1
XP_016876663.1
4
2212
XM_017021175.1
XP_016876664.1
4
2330
XM_017021176.1
XP_016876665.1
9
2157
XM_017021177.1
XP_016876666.1
9
2146
XM_017021178.1
XP_016876667.1
7
1984
XM_017021179.1
XP_016876668.1
6
1887
XM_017021180.1
XP_016876669.1
10
2199
XM_017021181.1
XP_016876670.1
8
2104
XM_017021182.1
XP_016876671.1
7
2037
XM_017021183.1
XP_016876672.1
5
1845
XM_017021184.1
XP_016876673.1
9
2157
XM_017021185.1
XP_016876674.1
2
2185
XM_017021186.1
XP_016876675.1
8
2104
XM_017021187.1
XP_016876676.1
6
1970
XM_017021188.1
XP_016876677.1
5
1845
XM_017021189.1
XP_016876678.1
7
1954
XM_017021190.1
XP_016876679.1
8
2104
XM_017021191.1
XP_016876680.1
6
1912
XM_017021192.1
XP_016876681.1
5
1845
XM_017021193.1
XP_016876682.1
5
1843
XM_017021194.1
XP_016876683.1
8
2104
XM_017021195.1
XP_016876684.1
8
2104
XM_017021196.1
XP_016876685.1
8
2104
GenBank mRNA
AB007900.2
-
1
1320
AF090989.1
-
2
1583
AF090990.1
-
2
1583
AK122930.1
-
2
2185
BC146777.1
-
1
1320
9007226
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
actin cytoskeleton reorganization
regulation of GTPase activity
ephrin receptor signaling pathway
regulation of synaptic plasticity
regulation of axonogenesis
regulation of small GTPase mediated signal transduction
regulation of dendritic spine morphogenesis
activation of GTPase activity
molecular_function
GTPase activator activity
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0