Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GAPVD1 SIRNA Assays ›
Gene Symbol:
GAPVD1
Gene Name:
GTPase activating protein and VPS9 domains 1
Gene Aliases:
GAPEX5, GAPex-5, RAP6
Chromosome Location:
Chr.9: 125261794 - 125365011 on Build GRCh38
UniGene ID:
Hs.495134
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS119837
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GAPVD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282679.1
NP_001269608.1
12, 13
2387
NM_001282680.1
NP_001269609.1
12, 13
2387
NM_001282681.1
NP_001269610.1
9, 10
1992
NM_015635.3
NP_056450.2
10, 11
2221
XM_005251901.3
XP_005251958.1
11, 12
2251
XM_005251904.3
XP_005251961.1
11, 12
2248
XM_006717044.3
XP_006717107.1
11, 12
2173
XM_011518499.2
XP_011516801.1
11, 12
2265
XM_011518500.2
XP_011516802.1
12, 13
2382
XM_011518502.2
XP_011516804.1
11, 12
2193
XM_011518506.2
XP_011516808.1
11, 12
2190
XM_011518507.2
XP_011516809.1
11, 12
2270
XM_017014600.1
XP_016870089.1
10, 11
2075
XM_017014601.1
XP_016870090.1
10, 11
2178
XM_017014602.1
XP_016870091.1
12, 13
2398
XM_017014603.1
XP_016870092.1
12, 13
2345
XM_017014604.1
XP_016870093.1
13, 14
2447
XM_017014605.1
XP_016870094.1
10, 11
2178
XM_017014606.1
XP_016870095.1
12, 13
2292
XM_017014607.1
XP_016870096.1
12, 13
2301
XM_017014608.1
XP_016870097.1
10, 11
2111
XM_017014609.1
XP_016870098.1
12, 13
2360
XM_017014610.1
XP_016870099.1
10, 11
2178
XM_017014611.1
XP_016870100.1
11, 12
2171
GenBank mRNA
AB040954.3
-
12, 13
2313
AK000126.1
-
8, 9
1611
AK023841.1
-
3, 4
336
AK027049.1
-
8, 9
1073
AK056493.1
-
10, 11
2183
AK294542.1
-
10, 11
2221
AK296381.1
-
11, 12
2311
BC010027.2
-
9, 10
2060
BC021119.1
-
8, 9
1712
BC114937.1
-
12, 13
2270
40083103
BC114962.1
-
10, 11
2106
40083104
DQ233254.1
-
9, 10
1960
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
signal transduction
positive regulation of GTPase activity
regulation of protein transport
cell-cell adhesion
guanyl-nucleotide exchange factor activity
GTPase activator activity
GTPase activating protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0